Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01502048_m1
          See other TSGA10IP GE Assays ›
          Gene Symbol:
          Tsga10ip
          Gene Name:
          testis specific 10 interacting protein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 220772592 - 220786931 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01502048_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tsga10ip
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004278.3 NP_001004278.2 5 - 6 1450 65
          NM_001270736.1 NP_001257665.1 6 - 7 1375 65
          NM_001270737.1 NP_001257666.1 5 - 6 1324 65
          XM_006230847.2 XP_006230909.1 6 - 7 1650 65
          XM_008760155.1 XP_008758377.1 6 - 7 1660 65
          XM_008760156.1 XP_008758378.1 5 - 6 1608 65
          GenBank mRNA AY690667.1 - 5 - 6 1467 65
          BC097254.1 - 5 - 6 1305 65

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tsga10ip

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P132801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30874 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71128 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P132801 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P132801 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P132801 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P132801 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71518 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10727839 - -
          Affymetrix Rat Gene 1.0 ST Array 10727844 - -
          Affymetrix Rat Gene 1.0 ST Array 10727841 - -
          Affymetrix Rat Gene 1.0 ST Array 10727845 - -
          Affymetrix Rat Gene 1.0 ST Array 10727843 - -
          Affymetrix Rat Gene 1.0 ST Array 10727842 - -
          Affymetrix Rat Gene 1.0 ST Array 10727838 - -
          Affymetrix Rat Gene 1.0 ST Array 10727840 - -
          Affymetrix Rat Gene 1.0 ST Array 10727837 - -
          Affymetrix Rat Gene 1.1 ST Array 10727836 - -
          Affymetrix Rat Gene 1.0 ST Array 10727836 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21643216 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376525 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1530526 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc