Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01504192_m1
          See other EFR3A GE Assays ›
          Gene Symbol:
          Efr3a
          Gene Name:
          EFR3 homolog A (S. cerevisiae)
          Gene Aliases:
          RGD1305976
          Chromosome Location:
          Chr.7: 106535434 - 106616805 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01504192_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Efr3a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001130564.1 NP_001124036.1 5 - 6 697 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001226.rn.1

          Set Membership:

          Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P560655 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P523244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8689 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35903 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48313 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113935 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152641 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91434 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118836 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174212 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60145 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59615 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168863 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P560655 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P523244 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P523244 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P560655 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P523244 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P560655 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P782990 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P523244 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P21134 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P620665 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10896857 - -
          Affymetrix Rat Gene 1.0 ST Array 10896855 - -
          Affymetrix Rat Gene 1.0 ST Array 10896859 - -
          Affymetrix Rat Gene 1.0 ST Array 10896846 - -
          Affymetrix Rat Gene 1.0 ST Array 10896847 - -
          Affymetrix Rat Gene 1.0 ST Array 10896854 - -
          Affymetrix Rat Gene 1.0 ST Array 10896840 - -
          Affymetrix Rat Gene 1.0 ST Array 10896853 - -
          Affymetrix Rat Gene 1.0 ST Array 10896856 - -
          Affymetrix Rat Gene 1.0 ST Array 10896848 - -
          Affymetrix Rat Gene 1.0 ST Array 10896837 - -
          Affymetrix Rat Gene 1.0 ST Array 10896843 - -
          Affymetrix Rat Gene 1.0 ST Array 10896852 - -
          Affymetrix Rat Gene 1.0 ST Array 10896842 - -
          Affymetrix Rat Gene 1.0 ST Array 10896850 - -
          Affymetrix Rat Gene 1.0 ST Array 10896845 - -
          Affymetrix Rat Gene 1.0 ST Array 10896839 - -
          Affymetrix Rat Gene 1.0 ST Array 10896851 - -
          Affymetrix Rat Gene 1.0 ST Array 10896849 - -
          Affymetrix Rat Gene 1.0 ST Array 10896844 - -
          Affymetrix Rat Gene 1.0 ST Array 10896838 - -
          Affymetrix Rat Gene 1.0 ST Array 10896841 - -
          Affymetrix Rat Gene 1.1 ST Array 10896836 - -
          Rat Genome 230 2.0 Array 1390550_at - -
          Rat Genome 230 2.0 Array 1389526_PM_at - -
          Rat Genome 230 2.0 Array 1374400_at - -
          Rat Genome 230 2.0 Array 1390550_PM_at - -
          Rat Genome 230 2.0 Array 1395739_at - -
          Rat Genome 230 2.0 Array 1389526_at - -
          Rat Genome 230 2.0 Array 1374400_PM_at - -
          Rat Genome 230 2.0 Array 1395739_PM_at - -
          Affymetrix HT Rat Focus Array 1395739_at - -
          Affymetrix HT Rat Focus Array 1390550_at - -
          Affymetrix HT Rat Focus Array 1389526_at - -
          Affymetrix HT Rat Focus Array 1374400_PM_at - -
          Affymetrix HT Rat Focus Array 1389526_PM_at - -
          Affymetrix HT Rat Focus Array 1395739_PM_at - -
          Affymetrix HT Rat Focus Array 1390550_PM_at - -
          Affymetrix HT Rat Focus Array 1374400_at - -
          Affymetrix Rat Clariom S Assay PSR0700008924.rn.1 TC0700001226.rn.1 RTA TC
          Rat Expression Set 230 1389526_at - -
          Rat Expression Set 230 1390550_PM_at - -
          Rat Expression Set 230 1374400_PM_at - -
          Rat Expression Set 230 1389526_PM_at - -
          Rat Expression Set 230 1390550_at - -
          Rat Expression Set 230 1374400_at - -
          Rat Expression Set 230 1395739_at - -
          Rat Expression Set 230 1395739_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10896836 - -
          Rat Genome U34 Set rc_AA955157_at - -
          Rat Genome U34 Set rc_AI179370_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21595834 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361911 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0