Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01504663_m1
          See other PDXP GE Assays ›
          Gene Symbol:
          Pdxp
          Gene Name:
          pyridoxal (pyridoxine, vitamin B6) phosphatase
          Gene Aliases:
          Rbp1
          Chromosome Location:
          Chr.7: 120140460 - 120145908 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01504663_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pdxp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001135819.1 NP_001129291.1 1 - 2 592 82
          GenBank mRNA AF318578.1 - 1 - 2 245 82
          BC166563.1 - 1 - 2 592 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001432.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          protein dephosphorylation
          regulation of mitotic nuclear division
          dephosphorylation
          positive regulation of actin filament depolymerization
          actin rod assembly
          pyridoxal phosphate catabolic process
          regulation of cytokinesis
          cellular response to ATP
          magnesium ion binding
          phosphoprotein phosphatase activity
          phosphatase activity
          heat shock protein binding
          pyridoxal phosphatase activity
          L-phosphoserine phosphatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P009530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515103 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136949 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P009530 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P009530 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P009530 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P504077 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74875 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10897528 - -
          Affymetrix Rat Gene 1.0 ST Array 10897525 - -
          Affymetrix Rat Gene 1.0 ST Array 10897526 - -
          Affymetrix Rat Gene 1.0 ST Array 10897527 - -
          Affymetrix Rat Gene 1.1 ST Array 10897524 - -
          Rat Genome 230 2.0 Array 1370938_PM_at - -
          Rat Genome 230 2.0 Array 1370938_at - -
          Affymetrix HT Rat Focus Array 1370938_at - -
          Affymetrix HT Rat Focus Array 1370938_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700010631.rn.1 TC0700001432.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700010633.rn.1 TC0700001432.rn.1 RTA TC
          Rat Expression Set 230 1370938_at - -
          Rat Expression Set 230 1370938_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10897524 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22321253 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0