Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01505018_g1
          See other NSMCE2 GE Assays ›
          Gene Symbol:
          Nsmce2
          Gene Name:
          non-SMC element 2, MMS21 homolog (S. cerevisiae)
          Gene Aliases:
          RGD1305156
          Chromosome Location:
          Chr.7: 99677295 - 99900765 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01505018_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nsmce2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024876.1 NP_001020047.1 5 - 6 597 102
          XM_008765505.1 XP_008763727.1 4 - 5 2757 102
          GenBank mRNA BC097477.1 - 5 - 6 597 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001178.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nsmce2

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cell cycle
          telomere maintenance via recombination
          double-strand break repair via homologous recombination
          double-strand break repair via nonhomologous end joining
          protein sumoylation
          positive regulation of maintenance of mitotic sister chromatid cohesion
          positive regulation of mitotic metaphase/anaphase transition
          cell division
          cellular senescence
          zinc ion binding
          ligase activity
          SUMO transferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P991459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311997 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12047 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142529 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105333 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72851 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180285 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33233 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178535 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81423 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163190 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146611 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94065 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107487 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1995 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74990 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115151 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162612 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161139 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27478 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145798 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47051 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51203 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162394 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119944 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92464 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147326 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129806 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79642 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30831 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39704 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169473 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175170 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P991459 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P991459 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P991459 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P991459 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73251 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83433 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14711 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53113 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7601 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31678 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83932 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49979 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49714 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21454 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95495 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25776 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97427 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61208 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36213 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68022 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10896789 - -
          Affymetrix Rat Gene 1.0 ST Array 10896786 - -
          Affymetrix Rat Gene 1.0 ST Array 10896787 - -
          Affymetrix Rat Gene 1.0 ST Array 10896788 - -
          Affymetrix Rat Gene 1.0 ST Array 10896791 - -
          Affymetrix Rat Gene 1.0 ST Array 10896790 - -
          Affymetrix Rat Gene 1.0 ST Array 10896792 - -
          Affymetrix Rat Gene 1.1 ST Array 10896785 - -
          Rat Genome 230 2.0 Array 1389011_at - -
          Rat Genome 230 2.0 Array 1389011_PM_at - -
          Affymetrix HT Rat Focus Array 1389011_at - -
          Affymetrix HT Rat Focus Array 1389011_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700008658.rn.1 TC0700001178.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700008653.rn.1 TC0700001178.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700008660.rn.1 TC0700001178.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700008654.rn.1 TC0700001178.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700008657.rn.1 TC0700001178.rn.1 RTA TC
          Rat Expression Set 230 1389011_at - -
          Rat Expression Set 230 1389011_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10896785 - -
          Rat Genome U34 Set rc_AA963322_f_at - -
          Rat Genome U34 Set rc_AI235480_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22278487 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352433 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline