Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01505100_g1
          See other RPL3 GE Assays ›
          Gene Symbol:
          Rpl3
          Gene Name:
          ribosomal protein L3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 121297365 - 121302739 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01505100_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpl3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_198753.2 NP_942048.1 2 - 3 223 67
          GenBank mRNA BC058494.1 - 2 - 3 226 67
          FQ209477.1 - 2 - 3 223 67
          FQ211542.1 - 2 - 3 223 67
          FQ211764.1 - 2 - 3 223 67
          FQ212671.1 - 2 - 3 223 67
          FQ213017.1 - 2 - 3 223 67
          FQ213135.1 - 2 - 3 225 67
          FQ214177.1 - 2 - 3 223 67
          FQ214350.1 - 2 - 3 223 67
          FQ214687.1 - 2 - 3 220 67
          FQ215305.1 - 2 - 3 223 67
          FQ218147.1 - 2 - 3 223 67
          FQ219083.1 - 2 - 3 223 67
          FQ219302.1 - 2 - 3 223 67
          FQ219523.1 - 2 - 3 222 67
          FQ220029.1 - 2 - 3 222 67
          FQ220099.1 - 2 - 3 223 67
          FQ220378.1 - 2 - 3 223 67
          FQ220408.1 - 2 - 3 219 67
          FQ220423.1 - 2 - 3 222 67
          FQ220830.1 - 2 - 3 223 67
          FQ222720.1 - 2 - 3 223 67
          FQ222993.1 - 2 - 3 223 67
          FQ225290.1 - 2 - 3 223 67
          FQ225405.1 - 2 - 3 223 67
          FQ225635.1 - 2 - 3 223 67
          FQ226331.1 - 2 - 3 223 67
          FQ227897.1 - 2 - 3 223 67
          FQ228916.1 - 2 - 3 223 67
          FQ229153.1 - 2 - 3 223 67
          FQ229505.1 - 2 - 3 223 67
          FQ229795.1 - 2 - 3 219 67
          FQ229869.1 - 2 - 3 223 67
          FQ231551.1 - 2 - 3 219 67
          FQ232038.1 - 2 - 3 223 67
          FQ232101.1 - 2 - 3 223 67
          FQ233196.1 - 2 - 3 223 67
          FQ234897.1 - 2 - 3 223 67
          FQ235165.1 - 2 - 3 223 67
          X62166.1 - 2 - 3 227 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700003524.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rpl3

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          cellular response to interleukin-4
          RNA binding
          structural constituent of ribosome

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR0700027801.rn.1 TC0700003524.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700027798.rn.1 TC0700003524.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-ztd5s:80/100.66.134.6:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0