Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01505208_m1
          See other CACNA1I GE Assays ›
          Gene Symbol:
          Cacna1i
          Gene Name:
          calcium channel, voltage-dependent, T type, alpha 1I subunit
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 121521787 - 121619300 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01505208_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cacna1i
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020084.3 NP_064469.3 27 - 28 4661 65
          GenBank mRNA AF086827.2 - 4926 65
          AF290214.4 - 27 - 28 4661 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001476.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cacna1i

          Gene Ontology Categories:

          Function(s) Process(es)

          neuronal action potential
          flagellated sperm motility
          sleep
          regulation of ion transmembrane transport
          calcium ion transport into cytosol
          calcium ion import
          calcium ion transmembrane transport
          membrane depolarization during action potential
          low voltage-gated calcium channel activity

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P017228 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P039842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73600 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4601 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82691 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2482 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96165 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20772 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104239 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46778 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135035 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134617 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43229 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69926 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23699 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P039842 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P017228 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P017228 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P039842 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P017228 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P039842 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P405083 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P405084 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49752 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78929 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95874 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85975 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90664 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46435 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90181 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10897787 - -
          Affymetrix Rat Gene 1.0 ST Array 10897783 - -
          Affymetrix Rat Gene 1.0 ST Array 10897768 - -
          Affymetrix Rat Gene 1.0 ST Array 10897793 - -
          Affymetrix Rat Gene 1.0 ST Array 10897792 - -
          Affymetrix Rat Gene 1.0 ST Array 10897785 - -
          Affymetrix Rat Gene 1.0 ST Array 10897771 - -
          Affymetrix Rat Gene 1.0 ST Array 10897794 - -
          Affymetrix Rat Gene 1.0 ST Array 10897779 - -
          Affymetrix Rat Gene 1.0 ST Array 10897777 - -
          Affymetrix Rat Gene 1.0 ST Array 10897784 - -
          Affymetrix Rat Gene 1.0 ST Array 10897786 - -
          Affymetrix Rat Gene 1.0 ST Array 10897773 - -
          Affymetrix Rat Gene 1.0 ST Array 10897776 - -
          Affymetrix Rat Gene 1.0 ST Array 10897769 - -
          Affymetrix Rat Gene 1.0 ST Array 10897791 - -
          Affymetrix Rat Gene 1.0 ST Array 10897770 - -
          Affymetrix Rat Gene 1.0 ST Array 10897775 - -
          Affymetrix Rat Gene 1.0 ST Array 10897798 - -
          Affymetrix Rat Gene 1.0 ST Array 10897795 - -
          Affymetrix Rat Gene 1.0 ST Array 10897781 - -
          Affymetrix Rat Gene 1.0 ST Array 10897790 - -
          Affymetrix Rat Gene 1.0 ST Array 10897782 - -
          Affymetrix Rat Gene 1.0 ST Array 10897788 - -
          Affymetrix Rat Gene 1.0 ST Array 10897800 - -
          Affymetrix Rat Gene 1.0 ST Array 10897772 - -
          Affymetrix Rat Gene 1.0 ST Array 10897767 - -
          Affymetrix Rat Gene 1.0 ST Array 10897766 - -
          Affymetrix Rat Gene 1.0 ST Array 10897796 - -
          Affymetrix Rat Gene 1.0 ST Array 10897778 - -
          Affymetrix Rat Gene 1.0 ST Array 10897780 - -
          Affymetrix Rat Gene 1.0 ST Array 10897797 - -
          Affymetrix Rat Gene 1.0 ST Array 10897774 - -
          Affymetrix Rat Gene 1.0 ST Array 10897789 - -
          Affymetrix Rat Gene 1.1 ST Array 10897764 - -
          Rat Genome 230 2.0 Array 1370641_s_at - -
          Rat Genome 230 2.0 Array 1369211_at - -
          Rat Genome 230 2.0 Array 1370641_PM_s_at - -
          Rat Genome 230 2.0 Array 1369211_PM_at - -
          Affymetrix HT Rat Focus Array 1369211_at - -
          Affymetrix HT Rat Focus Array 1369211_PM_at - -
          Affymetrix HT Rat Focus Array 1370641_s_at - -
          Affymetrix HT Rat Focus Array 1370641_PM_s_at - -
          Affymetrix Rat Clariom S Assay PSR0700011130.rn.1 TC0700001476.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700011129.rn.1 TC0700001476.rn.1 RTA TC
          Rat Expression Set 230 1369211_at - -
          Rat Expression Set 230 1369211_PM_at - -
          Rat Expression Set 230 1370641_PM_s_at - -
          Rat Expression Set 230 1370641_s_at - -
          Affymetrix Rat Gene 1.0 ST Array 10897764 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0