Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01505561_m1
          See other OSR2 GE Assays ›
          Gene Symbol:
          Osr2
          Gene Name:
          odd-skipped related transciption factor 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 74047779 - 74054946 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01505561_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Osr2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012118.1 NP_001012118.1 1 - 2 353 104
          XM_006241542.2 XP_006241604.1 1 - 2 394 104
          GenBank mRNA BC079211.1 - 1 - 2 353 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700000956.rn.1

          Additional Information:

          This assay is in the 5'UTR region of the BC079211.1, NM_001012118.1, XM_006241542.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Osr2

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          metanephros development
          mesonephros development
          chondrocyte differentiation
          transcription by RNA polymerase II
          positive regulation of cell population proliferation
          positive regulation of gene expression
          cell differentiation
          positive regulation of bone mineralization
          osteoblast proliferation
          embryonic forelimb morphogenesis
          embryonic hindlimb morphogenesis
          embryonic skeletal limb joint morphogenesis
          middle ear morphogenesis
          odontogenesis
          embryonic digit morphogenesis
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          embryonic skeletal system morphogenesis
          pronephros development
          positive regulation of epithelial cell proliferation
          roof of mouth development
          embryonic skeletal joint morphogenesis
          head development
          bone morphogenesis
          eyelid development in camera-type eye
          embryonic skeletal joint development
          positive regulation of gastrulation
          RNA polymerase II transcription regulatory region sequence-specific DNA binding
          DNA-binding transcription activator activity, RNA polymerase II-specific
          sequence-specific DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P441181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531508 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49971 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P441181 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P441181 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P441181 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P441181 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59540 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10896109 - -
          Affymetrix Rat Gene 1.0 ST Array 10896111 - -
          Affymetrix Rat Gene 1.0 ST Array 10896110 - -
          Affymetrix Rat Gene 1.0 ST Array 10896113 - -
          Affymetrix Rat Gene 1.1 ST Array 10896108 - -
          Rat Genome 230 2.0 Array 1380133_PM_at - -
          Rat Genome 230 2.0 Array 1380133_at - -
          Affymetrix HT Rat Focus Array 1380133_PM_at - -
          Affymetrix HT Rat Focus Array 1380133_at - -
          Affymetrix Rat Clariom S Assay PSR0700007261.rn.1 TC0700000956.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700007260.rn.1 TC0700000956.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700007258.rn.1 TC0700000956.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700007263.rn.1 TC0700000956.rn.1 RTA TC
          Rat Expression Set 230 1380133_PM_at - -
          Rat Expression Set 230 1380133_at - -
          Affymetrix Rat Gene 1.0 ST Array 10896108 - -
          Rat Genome U34 Set rc_AI231391_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20699846 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375304 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0