Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01505741_m1
          See other TTLL1 GE Assays ›
          Gene Symbol:
          Ttll1
          Gene Name:
          tubulin tyrosine ligase-like family, member 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 124338396 - 124367724 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01505741_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ttll1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012200.1 NP_001012200.1 8 - 9 1003 74
          XM_006242102.2 XP_006242164.1 8 - 9 1008 74
          XM_006242103.2 XP_006242165.1 8 - 9 1054 74
          XM_006242104.1 XP_006242166.1 7 - 8 1049 74
          XM_008765764.1 XP_008763986.1 7 - 8 960 74
          GenBank mRNA BC087669.1 - 8 - 9 1003 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700003598.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ttll1

          Gene Ontology Categories:

          Function(s) Process(es)

          epithelial cilium movement involved in extracellular fluid movement
          protein polyglutamylation
          axoneme assembly
          cilium organization
          cilium assembly
          ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P450486 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P633840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387301 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143017 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49505 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48812 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P450486 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P633840 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P633840 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P450486 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P450486 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P633840 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P450486 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P633840 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57692 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46272 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67872 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56877 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10898167 - -
          Affymetrix Rat Gene 1.0 ST Array 10898168 - -
          Affymetrix Rat Gene 1.0 ST Array 10898176 - -
          Affymetrix Rat Gene 1.0 ST Array 10898171 - -
          Affymetrix Rat Gene 1.0 ST Array 10898166 - -
          Affymetrix Rat Gene 1.0 ST Array 10898174 - -
          Affymetrix Rat Gene 1.0 ST Array 10898177 - -
          Affymetrix Rat Gene 1.0 ST Array 10898172 - -
          Affymetrix Rat Gene 1.0 ST Array 10898169 - -
          Affymetrix Rat Gene 1.0 ST Array 10898175 - -
          Affymetrix Rat Gene 1.0 ST Array 10898170 - -
          Affymetrix Rat Gene 1.0 ST Array 10898173 - -
          Affymetrix Rat Gene 1.1 ST Array 10898165 - -
          Rat Genome 230 2.0 Array 1383808_at - -
          Rat Genome 230 2.0 Array 1383808_PM_at - -
          Affymetrix HT Rat Focus Array 1383808_at - -
          Affymetrix HT Rat Focus Array 1383808_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700028493.rn.1 TC0700003598.rn.1 RTA TC
          Rat Expression Set 230 1383808_PM_at - -
          Rat Expression Set 230 1383808_at - -
          Affymetrix Rat Gene 1.0 ST Array 10898165 - -
          Rat Genome U34 Set rc_AA818423_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22128491 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375649 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650455 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376505 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-ffb8c786-92mpb:80/100.66.129.117:80.
          git-commit: 1a801765356e65be0b6ed8595ed79b52bb698fed
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0