Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01505881_m1
          See other STAT6 GE Assays ›
          Gene Symbol:
          Stat6
          Gene Name:
          signal transducer and activator of transcription 6, interleukin-4 induced
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 70946228 - 70963542 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01505881_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Stat6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001044250.1 NP_001037715.1 11 - 12 1291 55
          GenBank mRNA DQ462201.1 - 11 - 12 1291 55

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700000905.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          T-helper 1 cell lineage commitment
          negative regulation of type 2 immune response
          transcription, DNA-templated
          signal transduction
          cytokine-mediated signaling pathway
          mammary gland epithelial cell proliferation
          response to cytokine
          interleukin-4-mediated signaling pathway
          regulation of cell population proliferation
          positive regulation of transcription by RNA polymerase II
          positive regulation of isotype switching to IgE isotypes
          mammary gland morphogenesis
          cellular response to hydrogen peroxide
          cellular response to reactive nitrogen species
          RNA polymerase II core promoter sequence-specific DNA binding
          DNA binding
          DNA-binding transcription factor activity
          protein phosphatase binding
          identical protein binding
          sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P151804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892828 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87284 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56094 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P151804 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P151804 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P151804 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P151804 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98551 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91377 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10895952 - -
          Affymetrix Rat Gene 1.0 ST Array 10895935 - -
          Affymetrix Rat Gene 1.0 ST Array 10895937 - -
          Affymetrix Rat Gene 1.0 ST Array 10895955 - -
          Affymetrix Rat Gene 1.0 ST Array 10895939 - -
          Affymetrix Rat Gene 1.0 ST Array 10895941 - -
          Affymetrix Rat Gene 1.0 ST Array 10895950 - -
          Affymetrix Rat Gene 1.0 ST Array 10895944 - -
          Affymetrix Rat Gene 1.0 ST Array 10895942 - -
          Affymetrix Rat Gene 1.0 ST Array 10895945 - -
          Affymetrix Rat Gene 1.0 ST Array 10895957 - -
          Affymetrix Rat Gene 1.0 ST Array 10895956 - -
          Affymetrix Rat Gene 1.0 ST Array 10895943 - -
          Affymetrix Rat Gene 1.0 ST Array 10895936 - -
          Affymetrix Rat Gene 1.0 ST Array 10895947 - -
          Affymetrix Rat Gene 1.0 ST Array 10895940 - -
          Affymetrix Rat Gene 1.0 ST Array 10895954 - -
          Affymetrix Rat Gene 1.0 ST Array 10895946 - -
          Affymetrix Rat Gene 1.0 ST Array 10895934 - -
          Affymetrix Rat Gene 1.0 ST Array 10895938 - -
          Affymetrix Rat Gene 1.0 ST Array 10895953 - -
          Affymetrix Rat Gene 1.1 ST Array 10895932 - -
          Rat Genome 230 2.0 Array 1382350_at - -
          Rat Genome 230 2.0 Array 1391527_at - -
          Rat Genome 230 2.0 Array 1374925_at - -
          Rat Genome 230 2.0 Array 1374925_PM_at - -
          Rat Genome 230 2.0 Array 1391527_PM_at - -
          Rat Genome 230 2.0 Array 1382350_PM_at - -
          Affymetrix HT Rat Focus Array 1382350_PM_at - -
          Affymetrix HT Rat Focus Array 1382350_at - -
          Affymetrix Rat Clariom S Assay PSR0700006902.rn.1 TC0700000905.rn.1 RTA TC
          Rat Expression Set 230 1382350_PM_at - -
          Rat Expression Set 230 1382350_at - -
          Affymetrix Rat Gene 1.0 ST Array 10895932 - -
          Rat Genome U34 Set AF055292mRNA_at - -
          Rat Genome U34 Set AF055292mRNA_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20916779 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357297 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0