Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01508798_g1
          See other LRRC51 GE Assays ›
          Gene Symbol:
          Lrrc51
          Gene Name:
          leucine rich repeat containing 51
          Gene Aliases:
          RGD1565856
          Chromosome Location:
          Chr.1: 166997973 - 167017215 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01508798_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lrrc51
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106284.1 NP_001099754.1 5 - 6 687 141
          XM_006229870.2 XP_006229932.1 4 - 5 911 141
          XM_006229871.2 XP_006229933.1 4 - 5 798 141
          XM_006229872.2 XP_006229934.1 4 - 5 545 141
          XM_006229873.2 XP_006229935.1 4 - 5 587 141
          XM_006229874.2 XP_006229936.1 5 - 6 643 141
          XM_006229875.2 XP_006229937.1 4 - 5 513 141
          GenBank mRNA EU627092.1 - 4 - 5 556 141

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006170.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P22401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241889 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91113 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122810 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P22401 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P22401 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P22401 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P22401 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17989 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10724128 - -
          Affymetrix Rat Gene 1.0 ST Array 10724127 - -
          Affymetrix Rat Gene 1.0 ST Array 10724130 - -
          Affymetrix Rat Gene 1.0 ST Array 10724126 - -
          Affymetrix Rat Gene 1.0 ST Array 10724129 - -
          Affymetrix Rat Gene 1.1 ST Array 10724125 - -
          Rat Genome 230 2.0 Array 1398133_at - -
          Rat Genome 230 2.0 Array 1390667_at - -
          Rat Genome 230 2.0 Array 1398133_PM_at - -
          Rat Genome 230 2.0 Array 1385810_PM_at - -
          Rat Genome 230 2.0 Array 1385810_at - -
          Rat Genome 230 2.0 Array 1390667_PM_at - -
          Affymetrix HT Rat Focus Array 1390667_PM_at - -
          Affymetrix HT Rat Focus Array 1390667_at - -
          Affymetrix HT Rat Focus Array 1385810_PM_at - -
          Affymetrix HT Rat Focus Array 1385810_at - -
          Affymetrix Rat Clariom S Assay PSR0100049068.rn.1 TC0100006170.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100049069.rn.1 TC0100006170.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10724125 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21565200 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357535 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0