Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01510632_g1
          See other SULT1A1 GE Assays ›
          Gene Symbol:
          Sult1a1
          Gene Name:
          sulfotransferase family, cytosolic, 1A, phenol-preferring, member 1
          Gene Aliases:
          ASTIV, Mx-ST, PST-1, St1a1, Stm, Stp1, Sult1a3
          Chromosome Location:
          Chr.1: 198100586 - 198104106 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01510632_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sult1a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031834.1 NP_114022.1 7 - 8 844 80
          GenBank mRNA AF394783.1 - 6 - 7 767 80
          FQ209630.1 - 7 - 8 853 80
          FQ210886.1 - 7 - 8 849 80
          FQ214244.1 - 7 - 8 849 80
          FQ218459.1 - 7 - 8 849 80
          FQ218506.1 - 7 - 8 859 80
          FQ218658.1 - 7 - 8 848 80
          FQ219178.1 - 7 - 8 849 80
          FQ219948.1 - 7 - 8 849 80
          L19998.1 - 7 - 8 844 80
          X52883.1 - 7 - 8 832 80
          X68640.1 - 6 - 7 749 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006668.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sult1a1

          Gene Ontology Categories:

          Function(s) Process(es)

          catecholamine metabolic process
          xenobiotic metabolic process
          estrogen metabolic process
          regulation of blood pressure
          flavonoid metabolic process
          response to activity
          response to insecticide
          4-nitrophenol metabolic process
          response to glucocorticoid
          sulfation
          nucleotide binding
          aryl sulfotransferase activity
          sulfotransferase activity
          protein homodimerization activity
          flavonol 3-sulfotransferase activity
          steroid sulfotransferase activity
          3'-phosphoadenosine 5'-phosphosulfate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P011779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791283 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P011779 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P011779 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P011779 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P372501 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51408 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10725790 - -
          Affymetrix Rat Gene 1.0 ST Array 10725794 - -
          Affymetrix Rat Gene 1.0 ST Array 10725785 - -
          Affymetrix Rat Gene 1.0 ST Array 10725783 - -
          Affymetrix Rat Gene 1.0 ST Array 10725788 - -
          Affymetrix Rat Gene 1.0 ST Array 10725792 - -
          Affymetrix Rat Gene 1.0 ST Array 10725784 - -
          Affymetrix Rat Gene 1.0 ST Array 10725786 - -
          Affymetrix Rat Gene 1.0 ST Array 10725793 - -
          Affymetrix Rat Gene 1.0 ST Array 10725791 - -
          Affymetrix Rat Gene 1.1 ST Array 10725782 - -
          Rat Genome 230 2.0 Array 1370019_at - -
          Rat Genome 230 2.0 Array 1370019_PM_at - -
          Affymetrix HT Rat Focus Array 1370019_at - -
          Affymetrix HT Rat Focus Array 1370019_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100052326.rn.1 TC0100006668.rn.1 RTA TC
          Rat Expression Set 230 1370019_at - -
          Rat Expression Set 230 1370019_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10725782 - -
          Rat Genome U34 Set L19998_at - -
          Rat Genome U34 Set L19998_g_at - -
          Rat Genome U34 Set L19998_at - -
          Rat Genome U34 Set L19998_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21593259 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353876 - -
          RatRef-12 Expression BeadChip ILMN_52849 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0