Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qser1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139493.1 | NP_001132965.1 | 8 - 9 | 4802 | 95 | |
XM_006234646.2 | XP_006234708.1 | 8 - 9 | 4905 | 95 |
Set Membership: |
![]() ![]() ![]() ![]() |