Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01515653_g1
          See other CLCNKB GE Assays ›
          Gene Symbol:
          Clcnkb
          Gene Name:
          chloride channel, voltage-sensitive Kb
          Gene Aliases:
          ClC-K2L, Clck2, Clcnk1l
          Chromosome Location:
          Chr.5: 159950384 - 159973574 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01515653_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Clcnkb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173103.1 NP_775126.1 17 - 18 1954 91
          XM_006239320.2 XP_006239382.1 20 - 21 4826 91
          XM_008764293.1 XP_008762515.1 19 - 20 4494 91
          XM_008764294.1 XP_008762516.1 19 - 20 2760 91
          XM_008764295.1 XP_008762517.1 18 - 19 2905 91
          XM_008764296.1 XP_008762518.1 17 - 18 2783 91
          XM_008764297.1 XP_008762519.1 17 - 18 2765 91
          GenBank mRNA D26111.1 - 18 - 19 2008 91
          DQ437671.1 - 18 - 19 2058 91
          DQ437672.1 - 18 - 19 1951 91
          DQ437673.1 - 17 - 18 1929 91
          DQ437674.1 - 17 - 18 1893 91
          DQ437675.1 - 18 - 19 1901 91
          DQ437676.1 - 17 - 18 1879 91
          Z30663.1 - 17 - 18 1954 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500003957.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Clcnkb

          Gene Ontology Categories:

          Function(s) Process(es)

          chloride transport
          transepithelial chloride transport
          ion transmembrane transport
          regulation of ion transmembrane transport
          regulation of anion transport
          chloride transmembrane transport
          voltage-gated chloride channel activity
          chloride channel activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P029878 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P011544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274588 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26991 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P029878 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P011544 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P011544 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P029878 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P011544 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P029878 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P186883 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701825884 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56110 - -
          Agilent Rat cDNA Microarray (G4105A) 701825884 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10881097 - -
          Affymetrix Rat Gene 1.0 ST Array 10881096 - -
          Affymetrix Rat Gene 1.0 ST Array 10881090 - -
          Affymetrix Rat Gene 1.0 ST Array 10881086 - -
          Affymetrix Rat Gene 1.0 ST Array 10881092 - -
          Affymetrix Rat Gene 1.0 ST Array 10881088 - -
          Affymetrix Rat Gene 1.0 ST Array 10881094 - -
          Affymetrix Rat Gene 1.0 ST Array 10881085 - -
          Affymetrix Rat Gene 1.0 ST Array 10881098 - -
          Affymetrix Rat Gene 1.0 ST Array 10881099 - -
          Affymetrix Rat Gene 1.0 ST Array 10881091 - -
          Affymetrix Rat Gene 1.0 ST Array 10881095 - -
          Affymetrix Rat Gene 1.0 ST Array 10881103 - -
          Affymetrix Rat Gene 1.0 ST Array 10881089 - -
          Affymetrix Rat Gene 1.0 ST Array 10881104 - -
          Affymetrix Rat Gene 1.0 ST Array 10881101 - -
          Affymetrix Rat Gene 1.0 ST Array 10881100 - -
          Affymetrix Rat Gene 1.0 ST Array 10881105 - -
          Affymetrix Rat Gene 1.0 ST Array 10881087 - -
          Affymetrix Rat Gene 1.0 ST Array 10881093 - -
          Affymetrix Rat Gene 1.0 ST Array 10881102 - -
          Affymetrix Rat Gene 1.1 ST Array 10881084 - -
          Rat Genome 230 2.0 Array 1388175_at - -
          Rat Genome 230 2.0 Array 1388175_PM_at - -
          Affymetrix HT Rat Focus Array 1388175_at - -
          Affymetrix HT Rat Focus Array 1388175_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500029886.rn.1 TC0500003957.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500029885.rn.1 TC0500003957.rn.1 RTA TC
          Rat Expression Set 230 1388175_at - -
          Rat Expression Set 230 1388175_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10881084 - -
          Rat Genome U34 Set D26111_s_at - -
          Rat Genome U34 Set D26111_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21605427 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355888 - -
          RatRef-12 Expression BeadChip ILMN_59258 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0