Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01518088_m1
          See other NUMB GE Assays ›
          Gene Symbol:
          Numb
          Gene Name:
          numb homolog (Drosophila)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 107279917 - 107325345 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01518088_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Numb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133287.1 NP_579821.1 7 - 8 1065 69
          GenBank mRNA AB210108.1 - 7 - 8 1065 69
          DQ336702.1 - 8 - 9 1099 69
          DQ336703.1 - 7 - 8 1066 69

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Numb

          Gene Ontology Categories:

          Function(s) Process(es)

          nervous system development
          neuroblast proliferation
          axonogenesis
          cell population proliferation
          lateral ventricle development
          neuroblast division in subventricular zone
          cell differentiation
          positive regulation of cell migration
          positive regulation of polarized epithelial cell differentiation
          forebrain development
          adherens junction organization
          regulation of neuron differentiation
          negative regulation of Notch signaling pathway
          positive regulation of neurogenesis
          positive regulation of dendrite morphogenesis
          lung epithelial cell differentiation
          negative regulation of protein localization to plasma membrane
          protein binding
          beta-catenin binding
          alpha-catenin binding
          cadherin binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P040975 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P17757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230650 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162376 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146142 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152300 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167925 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11592 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127902 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86495 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90984 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123988 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63519 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124985 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63956 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P040975 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P17757 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P040975 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P17757 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P17757 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P040975 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P538526 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1032362 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12241 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13032 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95138 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7405 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103051 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84718 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26381 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46461 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10891045 - -
          Affymetrix Rat Gene 1.0 ST Array 10891049 - -
          Affymetrix Rat Gene 1.0 ST Array 10891042 - -
          Affymetrix Rat Gene 1.0 ST Array 10891044 - -
          Affymetrix Rat Gene 1.0 ST Array 10891043 - -
          Affymetrix Rat Gene 1.0 ST Array 10891040 - -
          Affymetrix Rat Gene 1.0 ST Array 10891048 - -
          Affymetrix Rat Gene 1.0 ST Array 10891046 - -
          Affymetrix Rat Gene 1.0 ST Array 10891047 - -
          Affymetrix Rat Gene 1.0 ST Array 10891041 - -
          Affymetrix Rat Gene 1.1 ST Array 10891039 - -
          Rat Genome 230 2.0 Array 1384750_PM_at - -
          Rat Genome 230 2.0 Array 1375536_PM_at - -
          Rat Genome 230 2.0 Array 1375536_at - -
          Rat Genome 230 2.0 Array 1384750_at - -
          Affymetrix HT Rat Focus Array 1375536_at - -
          Affymetrix HT Rat Focus Array 1384750_PM_at - -
          Affymetrix HT Rat Focus Array 1375536_PM_at - -
          Affymetrix HT Rat Focus Array 1384750_at - -
          Rat Expression Set 230 1375536_at - -
          Rat Expression Set 230 1375536_PM_at - -
          Rat Expression Set 230 1384750_at - -
          Rat Expression Set 230 1384750_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10891039 - -
          Rat Genome U34 Set rc_AI013849_at - -
          Rat Genome U34 Set rc_AI177848_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353560 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0