Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01518205_m1
          See other TTC7B GE Assays ›
          Gene Symbol:
          Ttc7b
          Gene Name:
          tetratricopeptide repeat domain 7B
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 124316444 - 124528058 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01518205_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ttc7b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108719.2 NP_001102189.1 13 - 14 1543 58
          XM_008764816.1 XP_008763038.1 19 - 20 2253 58
          GenBank mRNA BC168996.1 - 13 - 14 1543 58
          FQ210625.1 - 16 - 17 1722 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600003021.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P061345 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P477625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364248 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4545 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61068 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130402 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170886 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123315 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154056 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20049 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1437 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137171 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135003 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62672 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89925 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65269 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94208 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P061345 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P477625 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P061345 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P477625 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P477625 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P061345 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P356711 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45129 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24407 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45601 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27340 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89628 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5077 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10891652 - -
          Affymetrix Rat Gene 1.0 ST Array 10891642 - -
          Affymetrix Rat Gene 1.0 ST Array 10891649 - -
          Affymetrix Rat Gene 1.0 ST Array 10891631 - -
          Affymetrix Rat Gene 1.0 ST Array 10891635 - -
          Affymetrix Rat Gene 1.0 ST Array 10891651 - -
          Affymetrix Rat Gene 1.0 ST Array 10891634 - -
          Affymetrix Rat Gene 1.0 ST Array 10891650 - -
          Affymetrix Rat Gene 1.0 ST Array 10891644 - -
          Affymetrix Rat Gene 1.0 ST Array 10891639 - -
          Affymetrix Rat Gene 1.0 ST Array 10891647 - -
          Affymetrix Rat Gene 1.0 ST Array 10891645 - -
          Affymetrix Rat Gene 1.0 ST Array 10891648 - -
          Affymetrix Rat Gene 1.1 ST Array 10891630 - -
          Rat Genome 230 2.0 Array 1396631_PM_at - -
          Rat Genome 230 2.0 Array 1396631_at - -
          Rat Genome 230 2.0 Array 1376035_at - -
          Rat Genome 230 2.0 Array 1376035_PM_at - -
          Affymetrix HT Rat Focus Array 1376035_at - -
          Affymetrix HT Rat Focus Array 1376035_PM_at - -
          Affymetrix HT Rat Focus Array 1396631_PM_at - -
          Affymetrix HT Rat Focus Array 1396631_at - -
          Affymetrix Rat Clariom S Assay PSR0600021285.rn.1 TC0600003021.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600021284.rn.1 TC0600003021.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600021283.rn.1 TC0600003021.rn.1 RTA TC
          Rat Expression Set 230 1376035_at - -
          Rat Expression Set 230 1376035_PM_at - -
          Rat Expression Set 230 1396631_at - -
          Rat Expression Set 230 1396631_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10891630 - -
          Rat Genome U34 Set rc_AA901357_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22013226 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362590 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0