Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01518412_m1
          See other SERPINA10 GE Assays ›
          Gene Symbol:
          Serpina10 LOC100909524
          Gene Name:
          serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 10
          protein Z-dependent protease inhibitor-like
          Gene Aliases:
          Rasp1
          -
          Chromosome Location:
          Chr.6: 127500014 - 127508470 on Build Rnor_6.0
          Chromosome Location:
          Chr.6: 129004249 - 129012705 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01518412_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serpina10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133617.2 NP_598301.2 4 - 5 1248 66
          GenBank mRNA BC088113.1 - 4 - 5 1248 66
          U55765.1 - 3 - 4 1160 66
          Gene Symbol: LOC100909524
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003750227.3 XP_003750275.1 4 - 5 1266 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600003070.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Serpina10
          Click here to find siRNAs for Gene LOC100909524

          Gene Ontology Categories:

          Function(s) Process(es)

          blood coagulation
          negative regulation of endopeptidase activity
          tissue regeneration
          regulation of acrosome reaction
          serine-type endopeptidase inhibitor activity
          heparin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P083369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417716 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71047 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12512 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P083369 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P083369 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P083369 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P543096 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35966 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51191 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10891919 - -
          Affymetrix Rat Gene 1.0 ST Array 10891921 - -
          Affymetrix Rat Gene 1.0 ST Array 10891918 - -
          Affymetrix Rat Gene 1.0 ST Array 10891917 - -
          Affymetrix Rat Gene 1.0 ST Array 10891920 - -
          Affymetrix Rat Gene 1.1 ST Array 10891916 - -
          Rat Genome 230 2.0 Array 1368790_at - -
          Rat Genome 230 2.0 Array 1368790_PM_at - -
          Affymetrix HT Rat Focus Array 1368790_at - -
          Affymetrix HT Rat Focus Array 1368790_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0600021851.rn.1 TC0600003070.rn.1 RTA TC
          Rat Expression Set 230 1368790_at - -
          Rat Expression Set 230 1368790_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10891916 - -
          Rat Genome U34 Set U55765_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21846151 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354651 - -
          RatRef-12 Expression BeadChip ILMN_53532 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0