Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01519439_mH
          See other PIGV GE Assays ›
          Gene Symbol:
          Pigv
          Gene Name:
          phosphatidylinositol glycan anchor biosynthesis, class V
          Gene Aliases:
          RGD1309526
          Chromosome Location:
          Chr.5: 151886132 - 151899030 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01519439_mH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pigv
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001010966.1 NP_001010966.1 3 - 4 1569 64
          XM_006239051.2 XP_006239113.1 1 - 2 2334 64
          XM_006239055.2 XP_006239117.1 3 - 4 1315 64
          XM_008764159.1 XP_008762381.1 3 - 4 2748 64
          XM_008764160.1 XP_008762382.1 3 - 4 1292 64
          GenBank mRNA AB196341.1 - 3 - 4 1569 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500003784.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pigv

          Gene Ontology Categories:

          Function(s) Process(es)

          GPI anchor biosynthetic process
          mannosylation
          mannosyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P175875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352839 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5681 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P175875 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P175875 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P175875 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P175875 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7824 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10872560 - -
          Affymetrix Rat Gene 1.0 ST Array 10872558 - -
          Affymetrix Rat Gene 1.0 ST Array 10872556 - -
          Affymetrix Rat Gene 1.0 ST Array 10872557 - -
          Affymetrix Rat Gene 1.0 ST Array 10872559 - -
          Affymetrix Rat Gene 1.0 ST Array 10872555 - -
          Affymetrix Rat Gene 1.1 ST Array 10872554 - -
          Rat Genome 230 2.0 Array 1391664_PM_at - -
          Rat Genome 230 2.0 Array 1391664_at - -
          Affymetrix HT Rat Focus Array 1391664_at - -
          Affymetrix HT Rat Focus Array 1391664_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500028297.rn.1 TC0500003784.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500028307.rn.1 TC0500003784.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500028318.rn.1 TC0500003784.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500028319.rn.1 TC0500003784.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500028308.rn.1 TC0500003784.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500028300.rn.1 TC0500003784.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500028311.rn.1 TC0500003784.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500028301.rn.1 TC0500003784.rn.1 RTA TC
          Rat Expression Set 230 1391664_at - -
          Rat Expression Set 230 1391664_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10872554 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371306 - -
          RatRef-12 Expression BeadChip ILMN_57982 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0