Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01519707_g1
          See other CLCNKA GE Assays ›
          Gene Symbol:
          Clcnka
          Gene Name:
          chloride channel, voltage-sensitive Ka
          Gene Aliases:
          Clcnk1
          Chromosome Location:
          Chr.5: 159931497 - 159946483 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01519707_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Clcnka
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053327.1 NP_445779.1 13 - 14 1339 75
          XM_008764291.1 XP_008762513.1 13 - 14 1455 75
          XM_008764292.1 XP_008762514.1 13 - 14 1425 75
          GenBank mRNA BC081761.1 - 13 - 14 1405 75
          D13927.1 - 13 - 14 1339 75
          Z34291.1 - 12 - 13 1296 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500003956.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Clcnka

          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          chloride transport
          body fluid secretion
          response to pH
          response to water deprivation
          ion transmembrane transport
          regulation of ion transmembrane transport
          regulation of anion transport
          regulation of body fluid levels
          response to calcium ion
          renal inner medulla development
          chloride transmembrane transport
          voltage-gated ion channel activity
          voltage-gated chloride channel activity
          chloride channel activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P105014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757288 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129119 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P105014 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P105014 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P105014 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P667829 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51807 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70964 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10881070 - -
          Affymetrix Rat Gene 1.0 ST Array 10881073 - -
          Affymetrix Rat Gene 1.0 ST Array 10881078 - -
          Affymetrix Rat Gene 1.0 ST Array 10881080 - -
          Affymetrix Rat Gene 1.0 ST Array 10881077 - -
          Affymetrix Rat Gene 1.0 ST Array 10881064 - -
          Affymetrix Rat Gene 1.0 ST Array 10881074 - -
          Affymetrix Rat Gene 1.0 ST Array 10881081 - -
          Affymetrix Rat Gene 1.0 ST Array 10881066 - -
          Affymetrix Rat Gene 1.0 ST Array 10881079 - -
          Affymetrix Rat Gene 1.0 ST Array 10881082 - -
          Affymetrix Rat Gene 1.0 ST Array 10881067 - -
          Affymetrix Rat Gene 1.0 ST Array 10881068 - -
          Affymetrix Rat Gene 1.0 ST Array 10881072 - -
          Affymetrix Rat Gene 1.0 ST Array 10881075 - -
          Affymetrix Rat Gene 1.0 ST Array 10881083 - -
          Affymetrix Rat Gene 1.0 ST Array 10881076 - -
          Affymetrix Rat Gene 1.0 ST Array 10881065 - -
          Affymetrix Rat Gene 1.0 ST Array 10881069 - -
          Affymetrix Rat Gene 1.0 ST Array 10881071 - -
          Affymetrix Rat Gene 1.1 ST Array 10881063 - -
          Rat Genome 230 2.0 Array 1368765_at - -
          Rat Genome 230 2.0 Array 1368765_PM_at - -
          Affymetrix HT Rat Focus Array 1368765_PM_at - -
          Affymetrix HT Rat Focus Array 1368765_at - -
          Affymetrix Rat Clariom S Assay PSR0500029861.rn.1 TC0500003956.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500029859.rn.1 TC0500003956.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500029864.rn.1 TC0500003956.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500029856.rn.1 TC0500003956.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500029862.rn.1 TC0500003956.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500029863.rn.1 TC0500003956.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500029865.rn.1 TC0500003956.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500029855.rn.1 TC0500003956.rn.1 RTA TC
          Rat Expression Set 230 1368765_PM_at - -
          Rat Expression Set 230 1368765_at - -
          Affymetrix Rat Gene 1.0 ST Array 10881063 - -
          Rat Genome U34 Set D13927_at - -
          Rat Genome U34 Set D13927_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21259032 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366891 - -
          RatRef-12 Expression BeadChip ILMN_63263 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0