Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01521399_m1
          See other CLCA5 GE Assays ›
          Gene Symbol:
          Clca5
          Gene Name:
          chloride channel calcium activated 5
          Gene Aliases:
          Clca2
          Chromosome Location:
          Chr.2: 250954423 - 250982696 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01521399_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Clca5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107450.1 NP_001100920.1 11 - 12 2175 243

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          chloride transmembrane transport
          chloride channel activity
          ligand-gated ion channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P147300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298831 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167869 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15531 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133727 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P147300 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P147300 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P147300 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P147300 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10705 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10827221 - -
          Affymetrix Rat Gene 1.0 ST Array 10827218 - -
          Affymetrix Rat Gene 1.0 ST Array 10827225 - -
          Affymetrix Rat Gene 1.0 ST Array 10827229 - -
          Affymetrix Rat Gene 1.0 ST Array 10827228 - -
          Affymetrix Rat Gene 1.0 ST Array 10827227 - -
          Affymetrix Rat Gene 1.0 ST Array 10827217 - -
          Affymetrix Rat Gene 1.0 ST Array 10827226 - -
          Affymetrix Rat Gene 1.0 ST Array 10827224 - -
          Affymetrix Rat Gene 1.0 ST Array 10827222 - -
          Affymetrix Rat Gene 1.0 ST Array 10827220 - -
          Affymetrix Rat Gene 1.0 ST Array 10827219 - -
          Affymetrix Rat Gene 1.0 ST Array 10827230 - -
          Affymetrix Rat Gene 1.0 ST Array 10827223 - -
          Affymetrix Rat Gene 1.1 ST Array 10827216 - -
          Rat Genome 230 2.0 Array 1393147_at - -
          Rat Genome 230 2.0 Array 1393147_PM_at - -
          Affymetrix HT Rat Focus Array 1393147_at - -
          Affymetrix HT Rat Focus Array 1393147_PM_at - -
          Rat Expression Set 230 1393147_PM_at - -
          Rat Expression Set 230 1393147_at - -
          Affymetrix Rat Gene 1.0 ST Array 10827216 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21850080 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361466 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline