Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01522883_g1
          See other ELTD1 GE Assays ›
          Gene Symbol:
          Eltd1
          Gene Name:
          EGF, latrophilin and seven transmembrane domain containing 1
          Gene Aliases:
          Etl
          Chromosome Location:
          Chr.2: 256609787 - 256724642 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01522883_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eltd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022294.1 NP_071630.1 15 - 16 2390 181
          GenBank mRNA AF192401.1 - 15 - 16 2390 181
          AF192402.1 - 16 - 17 2624 181

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200002399.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Eltd1

          Gene Ontology Categories:

          Function(s) Process(es)

          G protein-coupled receptor signaling pathway
          G protein-coupled receptor activity
          calcium ion binding
          protein dimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P118447 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496510 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172562 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150515 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93334 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27466 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137135 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126967 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130165 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125777 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33501 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47643 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28296 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12205 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P118447 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12205 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P118447 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12205 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P118447 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12205 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700522643 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28754 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20485 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68822 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67032 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26688 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4028 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46388 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54225 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90374 - -
          Agilent Rat cDNA Microarray (G4105A) 700522643 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10819785 - -
          Affymetrix Rat Gene 1.0 ST Array 10819771 - -
          Affymetrix Rat Gene 1.0 ST Array 10819786 - -
          Affymetrix Rat Gene 1.0 ST Array 10819772 - -
          Affymetrix Rat Gene 1.0 ST Array 10819783 - -
          Affymetrix Rat Gene 1.0 ST Array 10819780 - -
          Affymetrix Rat Gene 1.0 ST Array 10819782 - -
          Affymetrix Rat Gene 1.0 ST Array 10819773 - -
          Affymetrix Rat Gene 1.0 ST Array 10819779 - -
          Affymetrix Rat Gene 1.0 ST Array 10819775 - -
          Affymetrix Rat Gene 1.0 ST Array 10819776 - -
          Affymetrix Rat Gene 1.0 ST Array 10819774 - -
          Affymetrix Rat Gene 1.0 ST Array 10819777 - -
          Affymetrix Rat Gene 1.0 ST Array 10819781 - -
          Affymetrix Rat Gene 1.0 ST Array 10819778 - -
          Affymetrix Rat Gene 1.0 ST Array 10819784 - -
          Affymetrix Rat Gene 1.1 ST Array 10819770 - -
          Rat Genome 230 2.0 Array 1387547_PM_a_at - -
          Rat Genome 230 2.0 Array 1387547_a_at - -
          Affymetrix HT Rat Focus Array 1387547_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387547_a_at - -
          Affymetrix Rat Clariom S Assay PSR0200016844.rn.1 TC0200002399.rn.1 RTA TC
          Rat Expression Set 230 1387547_PM_a_at - -
          Rat Expression Set 230 1387547_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10819770 - -
          Rat Genome U34 Set rc_AI237194_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21801833 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369759 - -
          RatRef-12 Expression BeadChip ILMN_66456 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline