Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01525076_g1
          See other PLP2 GE Assays ›
          Gene Symbol:
          Plp2
          Gene Name:
          proteolipid protein 2 (colonic epithelium-enriched)
          Gene Aliases:
          A4-LSB
          Chromosome Location:
          Chr.X: 15679254 - 15682653 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01525076_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Plp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207601.1 NP_997484.1 2 - 3 333 68
          GenBank mRNA BC061844.1 - 2 - 3 333 68
          FQ216622.1 - 2 - 3 369 68
          FQ220564.1 - 2 - 3 372 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00000147.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Plp2

          Gene Ontology Categories:

          Process(es)

          chemokine binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711812 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11370 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11370 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11370 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11370 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10932537 - -
          Affymetrix Rat Gene 1.0 ST Array 10932535 - -
          Affymetrix Rat Gene 1.0 ST Array 10923128 - -
          Affymetrix Rat Gene 1.0 ST Array 10932536 - -
          Affymetrix Rat Gene 1.0 ST Array 10932538 - -
          Affymetrix Rat Gene 1.1 ST Array 10923127 - -
          Affymetrix Rat Gene 1.1 ST Array 10932534 - -
          Rat Genome 230 2.0 Array 1388393_at - -
          Rat Genome 230 2.0 Array 1388393_PM_at - -
          Affymetrix HT Rat Focus Array 1388393_PM_at - -
          Affymetrix HT Rat Focus Array 1388393_at - -
          Affymetrix Rat Clariom S Assay PSR0X00001166.rn.1 TC0X00000147.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00001163.rn.1 TC0X00000147.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00001164.rn.1 TC0X00000147.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00001165.rn.1 TC0X00000147.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00001167.rn.1 TC0X00000147.rn.1 RTA TC
          Rat Expression Set 230 1388393_at - -
          Rat Expression Set 230 1388393_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10923127 - -
          Affymetrix Rat Gene 1.0 ST Array 10932534 - -
          Rat Genome U34 Set rc_AI013740_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21521325 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349466 - -
          RatRef-12 Expression BeadChip ILMN_52244 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-ztd5s:80/100.66.134.6:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0