Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egfl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004070.2 | NP_001004070.1 | 6 - 7 | 684 | 97 | |
NM_001165880.1 | NP_001159352.1 | 7 - 8 | 832 | 97 | ||
XM_006255977.2 | XP_006256039.1 | 7 - 8 | 1553 | 97 | ||
XM_006255978.2 | XP_006256040.1 | 8 - 9 | 1439 | 97 | ||
XM_006255979.2 | XP_006256041.1 | 8 - 9 | 935 | 97 | ||
GenBank mRNA | BC127463.1 | - | 7 - 8 | 832 | 97 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |