Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01527126_g1
          See other LOC685179 GE Assays ›
          Gene Symbol:
          LOC685179
          Gene Name:
          similar to SWI/SNF-related matrix-associated actin-dependent regulator of chromatin c2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 2875902 - 2905463 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01527126_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC685179
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_001055795.5 XP_001055795.1 27 - 28 3636 144
          XM_002729767.4 XP_002729813.2 28 - 29 3729 144
          XM_006240805.2 XP_006240867.1 28 - 29 3726 144
          XM_006240806.2 XP_006240868.1 28 - 29 3708 144
          XM_006240807.2 XP_006240869.1 27 - 28 3633 144
          XM_006240808.2 XP_006240870.1 27 - 28 3615 144
          XM_008765050.1 XP_008763272.1 29 - 30 3459 144
          XM_008765051.1 XP_008763273.1 28 - 29 4573 144

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          nucleosome disassembly
          chromatin remodeling
          regulation of transcription by RNA polymerase II
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          DNA binding
          chromatin binding
          protein binding
          nucleosomal DNA binding

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P061098 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P084069 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P084069 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P061101 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P061098 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P084069 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P061098 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P061101 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P061098 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P084069 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P061101 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P833772 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P107972 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10893186 - -
          Affymetrix Rat Gene 1.0 ST Array 10893193 - -
          Affymetrix Rat Gene 1.0 ST Array 10893182 - -
          Affymetrix Rat Gene 1.0 ST Array 10893189 - -
          Affymetrix Rat Gene 1.0 ST Array 10893205 - -
          Affymetrix Rat Gene 1.0 ST Array 10893198 - -
          Affymetrix Rat Gene 1.0 ST Array 10893184 - -
          Affymetrix Rat Gene 1.0 ST Array 10893201 - -
          Affymetrix Rat Gene 1.0 ST Array 10893197 - -
          Affymetrix Rat Gene 1.0 ST Array 10893202 - -
          Affymetrix Rat Gene 1.0 ST Array 10893207 - -
          Affymetrix Rat Gene 1.0 ST Array 10893188 - -
          Affymetrix Rat Gene 1.0 ST Array 10893196 - -
          Affymetrix Rat Gene 1.0 ST Array 10893208 - -
          Affymetrix Rat Gene 1.0 ST Array 10893210 - -
          Affymetrix Rat Gene 1.0 ST Array 10893187 - -
          Affymetrix Rat Gene 1.0 ST Array 10893194 - -
          Affymetrix Rat Gene 1.0 ST Array 10893203 - -
          Affymetrix Rat Gene 1.0 ST Array 10893200 - -
          Affymetrix Rat Gene 1.0 ST Array 10893192 - -
          Affymetrix Rat Gene 1.0 ST Array 10893209 - -
          Affymetrix Rat Gene 1.0 ST Array 10893190 - -
          Affymetrix Rat Gene 1.0 ST Array 10893195 - -
          Affymetrix Rat Gene 1.0 ST Array 10893191 - -
          Affymetrix Rat Gene 1.0 ST Array 10893183 - -
          Affymetrix Rat Gene 1.0 ST Array 10893204 - -
          Affymetrix Rat Gene 1.0 ST Array 10893185 - -
          Affymetrix Rat Gene 1.0 ST Array 10893206 - -
          Affymetrix Rat Gene 1.1 ST Array 10893181 - -
          Rat Genome 230 2.0 Array 1379185_PM_at - -
          Rat Genome 230 2.0 Array 1383154_PM_at - -
          Rat Genome 230 2.0 Array 1381691_PM_at - -
          Rat Genome 230 2.0 Array 1379185_at - -
          Rat Genome 230 2.0 Array 1390043_PM_at - -
          Rat Genome 230 2.0 Array 1383154_at - -
          Rat Genome 230 2.0 Array 1381691_at - -
          Rat Genome 230 2.0 Array 1389978_PM_at - -
          Rat Genome 230 2.0 Array 1390043_at - -
          Rat Genome 230 2.0 Array 1371716_PM_at - -
          Rat Genome 230 2.0 Array 1389978_at - -
          Rat Genome 230 2.0 Array 1371716_at - -
          Affymetrix HT Rat Focus Array 1389978_at - -
          Affymetrix HT Rat Focus Array 1371716_at - -
          Affymetrix HT Rat Focus Array 1371716_PM_at - -
          Affymetrix HT Rat Focus Array 1389978_PM_at - -
          Affymetrix HT Rat Focus Array 1390043_PM_at - -
          Affymetrix HT Rat Focus Array 1390043_at - -
          Affymetrix HT Rat Focus Array 1383154_at - -
          Affymetrix HT Rat Focus Array 1379185_at - -
          Affymetrix HT Rat Focus Array 1379185_PM_at - -
          Affymetrix HT Rat Focus Array 1381691_PM_at - -
          Affymetrix HT Rat Focus Array 1381691_at - -
          Affymetrix HT Rat Focus Array 1383154_PM_at - -
          Rat Expression Set 230 1371716_at - -
          Rat Expression Set 230 1389978_PM_at - -
          Rat Expression Set 230 1371716_PM_at - -
          Rat Expression Set 230 1390043_at - -
          Rat Expression Set 230 1389978_at - -
          Rat Expression Set 230 1390043_PM_at - -
          Rat Expression Set 230 1383154_PM_at - -
          Rat Expression Set 230 1383154_at - -
          Rat Expression Set 230 1379185_at - -
          Rat Expression Set 230 1381691_PM_at - -
          Rat Expression Set 230 1381691_at - -
          Rat Expression Set 230 1379185_PM_at - -
          Rat Genome U34 Set rc_AI008360_at - -
          Rat Genome U34 Set rc_AI111407_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0