Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01527158_m1
          See other RAB5B GE Assays ›
          Gene Symbol:
          Rab5b
          Gene Name:
          RAB5B, member RAS oncogene family
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 3104530 - 3123377 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01527158_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rab5b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001079936.1 NP_001073405.1 5 - 6 796 113
          XM_006240750.2 XP_006240812.1 5 - 6 719 113
          XM_006240751.2 XP_006240813.1 6 - 7 817 113
          XM_006240752.2 XP_006240814.1 5 - 6 944 113
          GenBank mRNA BC093595.1 - 1 - 2 46 113
          BC129101.1 - 5 - 6 796 113
          FQ211489.1 - 5 - 6 934 113

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700002003.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          endosome organization
          metabolic process
          antigen processing and presentation
          regulation of endocytosis
          Rab protein signal transduction
          plasma membrane to endosome transport
          GTPase activity
          GTP binding
          GDP binding
          GTP-dependent protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P029820 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P795009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408906 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168010 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108876 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81796 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P795009 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P029820 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P795009 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P029820 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P795009 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P029820 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P147582 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35535 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25324 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10899893 - -
          Affymetrix Rat Gene 1.0 ST Array 10899895 - -
          Affymetrix Rat Gene 1.0 ST Array 10899891 - -
          Affymetrix Rat Gene 1.0 ST Array 10899892 - -
          Affymetrix Rat Gene 1.0 ST Array 10899898 - -
          Affymetrix Rat Gene 1.0 ST Array 10899899 - -
          Affymetrix Rat Gene 1.0 ST Array 10899896 - -
          Affymetrix Rat Gene 1.0 ST Array 10899897 - -
          Affymetrix Rat Gene 1.1 ST Array 10899890 - -
          Rat Genome 230 2.0 Array 1393288_at - -
          Rat Genome 230 2.0 Array 1388823_at - -
          Rat Genome 230 2.0 Array 1399153_PM_at - -
          Rat Genome 230 2.0 Array 1399153_at - -
          Rat Genome 230 2.0 Array 1392643_PM_at - -
          Rat Genome 230 2.0 Array 1393288_PM_at - -
          Rat Genome 230 2.0 Array 1392643_at - -
          Rat Genome 230 2.0 Array 1388823_PM_at - -
          Affymetrix HT Rat Focus Array 1392643_at - -
          Affymetrix HT Rat Focus Array 1393288_PM_at - -
          Affymetrix HT Rat Focus Array 1399153_at - -
          Affymetrix HT Rat Focus Array 1392643_PM_at - -
          Affymetrix HT Rat Focus Array 1399153_PM_at - -
          Affymetrix HT Rat Focus Array 1393288_at - -
          Affymetrix HT Rat Focus Array 1388823_at - -
          Affymetrix HT Rat Focus Array 1388823_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700015770.rn.1 TC0700002003.rn.1 RTA TC
          Rat Expression Set 230 1388823_PM_at - -
          Rat Expression Set 230 1399153_at - -
          Rat Expression Set 230 1399153_PM_at - -
          Rat Expression Set 230 1388823_at - -
          Rat Expression Set 230 1392643_PM_at - -
          Rat Expression Set 230 1392643_at - -
          Rat Expression Set 230 1393288_PM_at - -
          Rat Expression Set 230 1393288_at - -
          Affymetrix Rat Gene 1.0 ST Array 10899890 - -
          Rat Genome U34 Set rc_AA866364_at - -
          Rat Genome U34 Set rc_AA996576_at - -
          Rat Genome U34 Set rc_AI060295_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21249024 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364389 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0