Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01527240_m1
          See other LOC100911681 GE Assays ›
          Gene Symbol:
          LOC100911681 LOC100910942 LOC103690055 Spink6
          Gene Name:
          serine protease inhibitor Kazal-type 6-like
          serine protease inhibitor Kazal-type 6-like
          serine protease inhibitor Kazal-type 6-like
          serine peptidase inhibitor, Kazal type 6
          Gene Aliases:
          -
          -
          -
          Spink5l1
          Chromosome Location:
          Chr.18: 35249137 - 35261046 on Build Rnor_6.0
          Chromosome Location:
          Chr.2: 36064130 - 36076039 on Build Rnor_6.0
          Chromosome Location:
          Chr.18: 35452004 - 35461304 on Build Rnor_6.0
          Chromosome Location:
          Chr.X: 43033066 - 43044975 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          Assay ID Rn01527240_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100911681
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003751772.2 XP_003751820.1 4 - 5 267 131
          Gene Symbol: LOC100910942
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003749193.1 XP_003749241.1 4 - 5 267 131
          Gene Symbol: LOC103690055
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008772230.1 XP_008770452.1 294 131
          Gene Symbol: Spink6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008874.1 NP_001008874.1 4 - 5 267 131
          XM_003752044.1 XP_003752092.1 4 - 5 267 131

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC100911681
          Click here to find siRNAs for Gene LOC100910942
          Click here to find siRNAs for Gene LOC103690055
          Click here to find siRNAs for Gene Spink6

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          negative regulation of endopeptidase activity
          serine-type endopeptidase inhibitor activity
          peptidase activity

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P153027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62537 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96190 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P153027 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P153027 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P153027 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97663 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14243 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10801474 - -
          Affymetrix Rat Gene 1.0 ST Array 10801472 - -
          Affymetrix Rat Gene 1.0 ST Array 10801475 - -
          Affymetrix Rat Gene 1.0 ST Array 10801473 - -
          Affymetrix Rat Gene 1.1 ST Array 10801471 - -
          Affymetrix Rat Gene 1.0 ST Array 10801471 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0