Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01528781_m1
          See other NDUFA7 GE Assays ›
          Gene Symbol:
          Ndufa7
          Gene Name:
          NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 7
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 18683553 - 18696332 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01528781_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ndufa7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106772.2 NP_001100242.2 2 - 3 115 63
          GenBank mRNA BC157817.1 - 2 - 3 115 63
          FQ224753.1 - 2 - 3 158 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700000334.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          ATP synthesis coupled electron transport
          molecular_function
          NADH dehydrogenase (ubiquinone) activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P139878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690099 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165327 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P139878 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P139878 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P139878 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1017013 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36657 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36818 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10894392 - -
          Affymetrix Rat Gene 1.0 ST Array 10894393 - -
          Affymetrix Rat Gene 1.0 ST Array 10894394 - -
          Affymetrix Rat Gene 1.0 ST Array 10894391 - -
          Affymetrix Rat Gene 1.1 ST Array 10894390 - -
          Rat Genome 230 2.0 Array 1395045_at - -
          Rat Genome 230 2.0 Array 1375411_PM_at - -
          Rat Genome 230 2.0 Array 1375411_at - -
          Rat Genome 230 2.0 Array 1395045_PM_at - -
          Affymetrix HT Rat Focus Array 1395045_PM_at - -
          Affymetrix HT Rat Focus Array 1395045_at - -
          Affymetrix HT Rat Focus Array 1375411_PM_at - -
          Affymetrix HT Rat Focus Array 1375411_at - -
          Affymetrix Rat Clariom S Assay PSR0700003019.rn.1 TC0700000334.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700003017.rn.1 TC0700000334.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700003015.rn.1 TC0700000334.rn.1 RTA TC
          Rat Expression Set 230 1375411_at - -
          Rat Expression Set 230 1375411_PM_at - -
          Rat Expression Set 230 1395045_PM_at - -
          Rat Expression Set 230 1395045_at - -
          Affymetrix Rat Gene 1.0 ST Array 10894390 - -
          Rat Genome U34 Set rc_AI013031_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350848 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0