Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01528837_m1
          See other RTFDC1 GE Assays ›
          Gene Symbol:
          Rtfdc1
          Gene Name:
          replication termination factor 2 domain containing 1
          Gene Aliases:
          RGD1311072
          Chromosome Location:
          Chr.3: 170443895 - 170473456 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01528837_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rtfdc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033890.1 NP_001029062.1 8 - 9 804 77
          GenBank mRNA BC101880.1 - 8 - 9 804 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002476.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rtfdc1

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P080967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184748 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5039 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160579 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142770 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P080967 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P080967 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P080967 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1032908 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11468 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2184 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77599 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1748 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104977 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10842545 - -
          Affymetrix Rat Gene 1.0 ST Array 10842539 - -
          Affymetrix Rat Gene 1.0 ST Array 10842540 - -
          Affymetrix Rat Gene 1.0 ST Array 10842546 - -
          Affymetrix Rat Gene 1.0 ST Array 10842542 - -
          Affymetrix Rat Gene 1.0 ST Array 10842541 - -
          Affymetrix Rat Gene 1.0 ST Array 10842544 - -
          Affymetrix Rat Gene 1.0 ST Array 10842543 - -
          Affymetrix Rat Gene 1.0 ST Array 10842547 - -
          Affymetrix Rat Gene 1.1 ST Array 10842538 - -
          Rat Genome 230 2.0 Array 1398949_PM_at - -
          Rat Genome 230 2.0 Array 1398949_at - -
          Affymetrix HT Rat Focus Array 1398949_PM_at - -
          Affymetrix HT Rat Focus Array 1398949_at - -
          Affymetrix Rat Clariom S Assay PSR0300018256.rn.1 TC0300002476.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300018257.rn.1 TC0300002476.rn.1 RTA TC
          Rat Expression Set 230 1398949_PM_at - -
          Rat Expression Set 230 1398949_at - -
          Affymetrix Rat Gene 1.0 ST Array 10842538 - -
          Rat Genome U34 Set rc_AI069986_at - -
          Rat Genome U34 Set rc_AI237872_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21553580 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356493 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0