Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01529709_m1
          See other ABCD1 GE Assays ›
          Gene Symbol:
          Abcd1
          Gene Name:
          ATP-binding cassette, subfamily D (ALD), member 1
          Gene Aliases:
          RGD1562128
          Chromosome Location:
          Chr.X: 157074117 - 157095511 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01529709_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Abcd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108821.1 NP_001102291.1 4 - 5 1828 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00002648.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          fatty acid beta-oxidation
          peroxisome organization
          long-chain fatty acid import into peroxisome
          long-chain fatty acid catabolic process
          very long-chain fatty acid catabolic process
          transmembrane transport
          long-chain fatty acid transporter activity
          protein binding
          ATP binding
          ATP hydrolysis activity
          enzyme binding
          ATPase-coupled transmembrane transporter activity
          identical protein binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P133973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158122 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27017 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167014 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160019 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P133973 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P133973 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P133973 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P18970 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44368 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10935959 - -
          Affymetrix Rat Gene 1.0 ST Array 10935953 - -
          Affymetrix Rat Gene 1.0 ST Array 10935952 - -
          Affymetrix Rat Gene 1.0 ST Array 10935957 - -
          Affymetrix Rat Gene 1.0 ST Array 10935951 - -
          Affymetrix Rat Gene 1.0 ST Array 10935955 - -
          Affymetrix Rat Gene 1.0 ST Array 10935958 - -
          Affymetrix Rat Gene 1.0 ST Array 10935956 - -
          Affymetrix Rat Gene 1.0 ST Array 10935954 - -
          Affymetrix Rat Gene 1.0 ST Array 10935950 - -
          Affymetrix Rat Gene 1.1 ST Array 10935949 - -
          Rat Genome 230 2.0 Array 1376140_at - -
          Rat Genome 230 2.0 Array 1376140_PM_at - -
          Affymetrix HT Rat Focus Array 1376140_at - -
          Affymetrix HT Rat Focus Array 1376140_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0X00016571.rn.1 TC0X00002648.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00016570.rn.1 TC0X00002648.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00016572.rn.1 TC0X00002648.rn.1 RTA TC
          Rat Expression Set 230 1376140_at - -
          Rat Expression Set 230 1376140_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10935949 - -
          Rat Genome U34 Set rc_AI170676_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21382317 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0