Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01529783_m1
          See other ATP2B3 GE Assays ›
          Gene Symbol:
          Atp2b3
          Gene Name:
          ATPase, Ca++ transporting, plasma membrane 3
          Gene Aliases:
          Pmca3
          Chromosome Location:
          Chr.X: 157236400 - 157312028 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01529783_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atp2b3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133288.1 NP_579822.1 20 - 21 3803 101
          XM_008773619.1 XP_008771841.1 18 - 19 3214 101
          XM_008773620.1 XP_008771842.1 17 - 18 3172 101
          XM_008773621.1 XP_008771843.1 18 - 19 3214 101
          XM_008773622.1 XP_008771844.1 17 - 18 3172 101
          XM_008773623.1 XP_008771845.1 18 - 19 3214 101
          XM_008773624.1 XP_008771846.1 18 - 19 3214 101
          XM_008773625.1 XP_008771847.1 18 - 19 3214 101
          XM_008773626.1 XP_008771848.1 18 - 19 3214 101
          XM_008773627.1 XP_008771849.1 18 - 19 3271 101
          GenBank mRNA BC089969.1 - 2 - 3 284 101
          J05087.1 - 20 - 21 3803 101
          M96626.1 - 1 - 2 107 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00002651.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          neural retina development
          brain development
          metabolic process
          calcium ion transmembrane transport
          P-type calcium transporter activity
          protein binding
          calmodulin binding
          ATP binding
          PDZ domain binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P035496 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P012725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970529 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119333 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137163 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125122 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132970 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109333 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36384 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46729 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83504 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P035496 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P012725 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P012725 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P035496 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P012725 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P035496 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P659399 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P723936 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88587 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55497 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86500 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35130 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10935926 - -
          Affymetrix Rat Gene 1.0 ST Array 10935919 - -
          Affymetrix Rat Gene 1.0 ST Array 10935915 - -
          Affymetrix Rat Gene 1.0 ST Array 10935909 - -
          Affymetrix Rat Gene 1.0 ST Array 10935901 - -
          Affymetrix Rat Gene 1.0 ST Array 10935927 - -
          Affymetrix Rat Gene 1.0 ST Array 10935905 - -
          Affymetrix Rat Gene 1.0 ST Array 10935911 - -
          Affymetrix Rat Gene 1.0 ST Array 10935903 - -
          Affymetrix Rat Gene 1.0 ST Array 10935921 - -
          Affymetrix Rat Gene 1.0 ST Array 10935906 - -
          Affymetrix Rat Gene 1.0 ST Array 10935923 - -
          Affymetrix Rat Gene 1.0 ST Array 10935910 - -
          Affymetrix Rat Gene 1.0 ST Array 10935913 - -
          Affymetrix Rat Gene 1.0 ST Array 10935900 - -
          Affymetrix Rat Gene 1.0 ST Array 10935925 - -
          Affymetrix Rat Gene 1.0 ST Array 10935904 - -
          Affymetrix Rat Gene 1.0 ST Array 10935912 - -
          Affymetrix Rat Gene 1.0 ST Array 10935920 - -
          Affymetrix Rat Gene 1.0 ST Array 10935917 - -
          Affymetrix Rat Gene 1.0 ST Array 10935918 - -
          Affymetrix Rat Gene 1.0 ST Array 10935902 - -
          Affymetrix Rat Gene 1.0 ST Array 10935907 - -
          Affymetrix Rat Gene 1.0 ST Array 10935924 - -
          Affymetrix Rat Gene 1.0 ST Array 10935914 - -
          Affymetrix Rat Gene 1.1 ST Array 10935899 - -
          Rat Genome 230 2.0 Array 1388037_at - -
          Rat Genome 230 2.0 Array 1388237_s_at - -
          Rat Genome 230 2.0 Array 1371172_PM_at - -
          Rat Genome 230 2.0 Array 1371172_at - -
          Rat Genome 230 2.0 Array 1388037_PM_at - -
          Rat Genome 230 2.0 Array 1388237_PM_s_at - -
          Affymetrix HT Rat Focus Array 1388237_PM_s_at - -
          Affymetrix HT Rat Focus Array 1371172_PM_at - -
          Affymetrix HT Rat Focus Array 1388237_s_at - -
          Affymetrix HT Rat Focus Array 1388037_at - -
          Affymetrix HT Rat Focus Array 1388037_PM_at - -
          Affymetrix HT Rat Focus Array 1371172_at - -
          Affymetrix Rat Clariom S Assay PSR0X00016611.rn.1 TC0X00002651.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00016612.rn.1 TC0X00002651.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00016613.rn.1 TC0X00002651.rn.1 RTA TC
          Rat Expression Set 230 1388237_PM_s_at - -
          Rat Expression Set 230 1388037_PM_at - -
          Rat Expression Set 230 1388237_s_at - -
          Rat Expression Set 230 1371172_PM_at - -
          Rat Expression Set 230 1371172_at - -
          Rat Expression Set 230 1388037_at - -
          Affymetrix Rat Gene 1.0 ST Array 10935899 - -
          Rat Genome U34 Set M96626_at - -
          Rat Genome U34 Set J05087_at - -
          Rat Genome U34 Set M96626_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21419658 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352628 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0