Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01530347_g1
          See other TRIM13 GE Assays ›
          Gene Symbol:
          Trim13
          Gene Name:
          tripartite motif-containing 13
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 41927180 - 41936529 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01530347_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Trim13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012210.1 NP_001012210.1 2 - 3 119 137
          XM_006252137.2 XP_006252199.1 2 - 3 206 137
          GenBank mRNA BC088425.1 - 2 - 3 119 137

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500000626.rn.1

          Additional Information:

          This assay is in the 5'UTR region of the BC088425.1, NM_001012210.1, XM_006252137.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Trim13

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          response to gamma radiation
          positive regulation of cell death
          positive regulation of macroautophagy
          ubiquitin-dependent ERAD pathway
          negative regulation of viral transcription
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          proteasome-mediated ubiquitin-dependent protein catabolic process
          innate immune response
          positive regulation of NF-kappaB transcription factor activity
          protein autoubiquitination
          negative regulation of viral release from host cell
          ubiquitin-protein transferase activity
          zinc ion binding
          ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P109977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357400 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22326 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P109977 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P109977 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P109977 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P109977 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92011 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76323 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10780929 - -
          Affymetrix Rat Gene 1.0 ST Array 10780930 - -
          Affymetrix Rat Gene 1.1 ST Array 10780924 - -
          Affymetrix Rat Clariom S Assay PSR1500004391.rn.1 TC1500000626.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500004392.rn.1 TC1500000626.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10780924 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22155216 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376444 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376030 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0