Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01530665_m1
          See other MYZAP GE Assays ›
          Gene Symbol:
          Myzap
          Gene Name:
          myocardial zonula adherens protein
          Gene Aliases:
          Gcom1
          Chromosome Location:
          Chr.8: 78009019 - 78096411 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01530665_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Myzap
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014211.1 NP_001014233.1 2 - 3 168 101
          GenBank mRNA BC089899.1 - 2 - 3 168 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800002879.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Myzap

          Gene Ontology Categories:

          Function(s)

          intracellular signal transduction

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P144023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488954 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71867 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12133 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168998 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5123 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23325 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14696 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133426 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63459 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83816 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167203 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173867 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8426 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P144023 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P144023 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P144023 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P112852 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78165 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71240 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69891 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33654 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55040 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69687 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52695 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10918590 - -
          Affymetrix Rat Gene 1.0 ST Array 10918594 - -
          Affymetrix Rat Gene 1.0 ST Array 10918592 - -
          Affymetrix Rat Gene 1.0 ST Array 10918591 - -
          Affymetrix Rat Gene 1.0 ST Array 10918588 - -
          Affymetrix Rat Gene 1.0 ST Array 10918595 - -
          Affymetrix Rat Gene 1.0 ST Array 10918589 - -
          Affymetrix Rat Gene 1.0 ST Array 10918599 - -
          Affymetrix Rat Gene 1.0 ST Array 10918597 - -
          Affymetrix Rat Gene 1.0 ST Array 10918593 - -
          Affymetrix Rat Gene 1.0 ST Array 10918587 - -
          Affymetrix Rat Gene 1.0 ST Array 10918596 - -
          Affymetrix Rat Gene 1.0 ST Array 10918598 - -
          Affymetrix Rat Gene 1.1 ST Array 10918586 - -
          Rat Genome 230 2.0 Array 1374816_PM_at - -
          Rat Genome 230 2.0 Array 1374816_at - -
          Affymetrix HT Rat Focus Array 1374816_PM_at - -
          Affymetrix HT Rat Focus Array 1374816_at - -
          Affymetrix Rat Clariom S Assay PSR0800023285.rn.1 TC0800002879.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800023287.rn.1 TC0800002879.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800023295.rn.1 TC0800002879.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800023282.rn.1 TC0800002879.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800023293.rn.1 TC0800002879.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800023294.rn.1 TC0800002879.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800023296.rn.1 TC0800002879.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800023286.rn.1 TC0800002879.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800023283.rn.1 TC0800002879.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800023284.rn.1 TC0800002879.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800023291.rn.1 TC0800002879.rn.1 RTA TC
          Rat Expression Set 230 1374816_PM_at - -
          Rat Expression Set 230 1374816_at - -
          Affymetrix Rat Gene 1.0 ST Array 10918586 - -
          Rat Genome U34 Set rc_AI103939_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20876539 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0