Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01530908_m1
          See other LOC103689982 GE Assays ›
          Gene Symbol:
          LOC103689982 Mboat7
          Gene Name:
          lysophospholipid acyltransferase 7
          membrane bound O-acyltransferase domain containing 7
          Gene Aliases:
          -
          Leng4, RGD1306945
          Chromosome Location:
          Chr.1: 64031859 - 64046137 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 64100159 - 64114437 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01530908_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC103689982
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008758837.1 XP_008757059.1 6 - 7 1075 68
          Gene Symbol: Mboat7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001134978.1 NP_001128450.1 1075 68
          GenBank mRNA BC169089.1 - 1075 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100000705.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P054043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823553 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69900 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142835 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P054043 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P054043 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P054043 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P813095 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10703718 - -
          Affymetrix Rat Gene 1.0 ST Array 10703721 - -
          Affymetrix Rat Gene 1.0 ST Array 10703719 - -
          Affymetrix Rat Gene 1.0 ST Array 10703716 - -
          Affymetrix Rat Gene 1.0 ST Array 10703720 - -
          Affymetrix Rat Gene 1.0 ST Array 10703717 - -
          Affymetrix Rat Gene 1.0 ST Array 10703722 - -
          Affymetrix Rat Gene 1.1 ST Array 10703715 - -
          Rat Genome 230 2.0 Array 1372680_PM_at - -
          Rat Genome 230 2.0 Array 1372680_at - -
          Affymetrix HT Rat Focus Array 1372680_PM_at - -
          Affymetrix HT Rat Focus Array 1372680_at - -
          Affymetrix Rat Clariom S Assay PSR0100004590.rn.1 TC0100000705.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100004588.rn.1 TC0100000705.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100004592.rn.1 TC0100000705.rn.1 RTA TC
          Rat Expression Set 230 1372680_at - -
          Rat Expression Set 230 1372680_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10703715 - -
          Rat Genome U34 Set rc_AA894212_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354751 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0