Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01530912_g1
          See other LOC103689992 GE Assays ›
          Gene Symbol:
          LOC103689992 Rps9
          Gene Name:
          40S ribosomal protein S9
          ribosomal protein S9
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.1: 64018327 - 64021747 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 64086628 - 64090017 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01530912_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC103689992
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008758836.1 XP_008757058.1 346 59
          Gene Symbol: Rps9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031108.3 NP_112370.2 3 - 4 316 59
          GenBank mRNA BC060560.1 - 3 - 4 321 59
          FQ229992.1 - 3 - 4 276 59
          X66370.1 - 2 - 3 250 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100004640.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC103689992
          Click here to find siRNAs for Gene Rps9

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          biological_process
          positive regulation of translational fidelity
          molecular_function
          structural constituent of ribosome
          rRNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P037030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443156 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36366 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P037030 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P037030 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P037030 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P553665 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700814534 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702450649 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67349 - -
          Agilent Rat cDNA Microarray (G4105A) 702450649 - -
          Agilent Rat cDNA Microarray (G4105A) 700814534 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10718511 - -
          Affymetrix Rat Gene 1.0 ST Array 10718512 - -
          Affymetrix Rat Gene 1.0 ST Array 10940481 - -
          Affymetrix Rat Gene 1.0 ST Array 10932837 - -
          Affymetrix Rat Gene 1.0 ST Array 10718513 - -
          Affymetrix Rat Gene 1.1 ST Array 10718510 - -
          Affymetrix Rat Gene 1.1 ST Array 10940481 - -
          Affymetrix Rat Gene 1.1 ST Array 10932836 - -
          Rat Genome 230 2.0 Array 1387888_PM_at - -
          Rat Genome 230 2.0 Array 1387888_at - -
          Affymetrix HT Rat Focus Array 1387888_PM_at - -
          Affymetrix HT Rat Focus Array 1387888_at - -
          Affymetrix Rat Clariom S Assay PSR0100036580.rn.1 TC0100004640.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100036585.rn.1 TC0100004640.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100036587.rn.1 TC0100004640.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100036581.rn.1 TC0100004640.rn.1 RTA TC
          Rat Expression Set 230 1387888_PM_at - -
          Rat Expression Set 230 1387888_at - -
          Affymetrix Rat Gene 1.0 ST Array 10932836 - -
          Affymetrix Rat Gene 1.0 ST Array 10940481 - -
          Affymetrix Rat Gene 1.0 ST Array 10718510 - -
          Rat Genome U34 Set X66370_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20918319 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2038795 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2038796 - -
          RatRef-12 Expression BeadChip ILMN_59654 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0