Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01531509_g1
          See other ESAM GE Assays ›
          Gene Symbol:
          Esam
          Gene Name:
          endothelial cell adhesion molecule
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 39997875 - 40008802 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01531509_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Esam
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004245.1 NP_001004245.1 5 - 6 828 90
          GenBank mRNA BC079093.1 - 5 - 6 828 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800000413.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Esam

          Gene Ontology Categories:

          Function(s) Process(es)

          homophilic cell adhesion via plasma membrane adhesion molecules
          biological_process
          cell-cell adhesion
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P481816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959849 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91812 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P481816 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P481816 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P481816 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P481816 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79488 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10909075 - -
          Affymetrix Rat Gene 1.0 ST Array 10909078 - -
          Affymetrix Rat Gene 1.0 ST Array 10909076 - -
          Affymetrix Rat Gene 1.0 ST Array 10909074 - -
          Affymetrix Rat Gene 1.0 ST Array 10909079 - -
          Affymetrix Rat Gene 1.0 ST Array 10909073 - -
          Affymetrix Rat Gene 1.0 ST Array 10909077 - -
          Affymetrix Rat Gene 1.1 ST Array 10909072 - -
          Rat Genome 230 2.0 Array 1388703_at - -
          Rat Genome 230 2.0 Array 1388703_PM_at - -
          Affymetrix HT Rat Focus Array 1388703_PM_at - -
          Affymetrix HT Rat Focus Array 1388703_at - -
          Affymetrix Rat Clariom S Assay PSR0800002754.rn.1 TC0800000413.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800002755.rn.1 TC0800000413.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800002752.rn.1 TC0800000413.rn.1 RTA TC
          Rat Expression Set 230 1388703_PM_at - -
          Rat Expression Set 230 1388703_at - -
          Affymetrix Rat Gene 1.0 ST Array 10909072 - -
          Rat Genome U34 Set rc_AI234105_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21006072 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351676 - -
          RatRef-12 Expression BeadChip ILMN_54237 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline