Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01531866_m1
          See other COLEC12 GE Assays ›
          Gene Symbol:
          Colec12
          Gene Name:
          collectin sub-family member 12
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 867048 - 1052608 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01531866_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Colec12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025721.1 NP_001020892.1 7 - 8 2035 119
          GenBank mRNA BC097495.1 - 7 - 8 2035 119

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800000010.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Colec12

          Gene Ontology Categories:

          Function(s) Process(es)

          pattern recognition receptor signaling pathway
          phagocytosis, recognition
          immune response
          low-density lipoprotein particle binding
          carbohydrate binding
          pattern recognition receptor activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P020466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604501 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108850 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6844 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176705 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70016 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6160 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76722 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27984 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138666 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155636 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23813 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68729 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107962 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75254 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121860 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145696 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130325 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107164 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6245 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105596 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10377 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P020466 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P020466 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P020466 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P598365 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P288029 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74625 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43191 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80079 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62690 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86669 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68066 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82889 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49234 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69180 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63259 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70574 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44032 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8575 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16071 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25982 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58498 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10800094 - -
          Affymetrix Rat Gene 1.0 ST Array 10800092 - -
          Affymetrix Rat Gene 1.0 ST Array 10800096 - -
          Affymetrix Rat Gene 1.0 ST Array 10800093 - -
          Affymetrix Rat Gene 1.0 ST Array 10800090 - -
          Affymetrix Rat Gene 1.0 ST Array 10800097 - -
          Affymetrix Rat Gene 1.0 ST Array 10800095 - -
          Affymetrix Rat Gene 1.0 ST Array 10800089 - -
          Affymetrix Rat Gene 1.0 ST Array 10800091 - -
          Affymetrix Rat Gene 1.1 ST Array 10800088 - -
          Rat Genome 230 2.0 Array 1372818_at - -
          Rat Genome 230 2.0 Array 1372818_PM_at - -
          Affymetrix HT Rat Focus Array 1372818_PM_at - -
          Affymetrix HT Rat Focus Array 1372818_at - -
          Affymetrix Rat Clariom S Assay PSR1800000074.rn.1 TC1800000010.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800000073.rn.1 TC1800000010.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800000072.rn.1 TC1800000010.rn.1 RTA TC
          Rat Expression Set 230 1372818_at - -
          Rat Expression Set 230 1372818_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10800088 - -
          Rat Genome U34 Set rc_AI180192_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21337559 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359457 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0