Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01533363_g1
          See other KRT18 GE Assays ›
          Gene Symbol:
          Krt18
          Gene Name:
          keratin 18, type I
          Gene Aliases:
          Krt1-18
          Chromosome Location:
          Chr.7: 143629455 - 143633131 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01533363_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Krt18
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053976.1 NP_446428.1 6 - 7 1194 101
          GenBank mRNA BC091275.1 - 6 - 7 1194 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001888.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Krt18

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          tumor necrosis factor-mediated signaling pathway
          Golgi to plasma membrane CFTR protein transport
          negative regulation of apoptotic process
          intermediate filament cytoskeleton organization
          extrinsic apoptotic signaling pathway
          hepatocyte apoptotic process
          molecular_function
          RNA binding
          structural molecule activity
          scaffold protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P115506 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P115506 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P115506 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P115506 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P400494 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1025476 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700601460 - -
          Agilent Rat cDNA Microarray (G4105A) 700601460 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10940455 - -
          Affymetrix Rat Gene 1.0 ST Array 10745346 - -
          Affymetrix Rat Gene 1.0 ST Array 10813993 - -
          Affymetrix Rat Gene 1.0 ST Array 10813994 - -
          Affymetrix Rat Gene 1.1 ST Array 10813992 - -
          Affymetrix Rat Gene 1.1 ST Array 10745345 - -
          Affymetrix Rat Gene 1.1 ST Array 10940455 - -
          Rat Genome 230 2.0 Array 1388155_at - -
          Rat Genome 230 2.0 Array 1388155_PM_at - -
          Affymetrix HT Rat Focus Array 1388155_at - -
          Affymetrix HT Rat Focus Array 1388155_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700014649.rn.1 TC0700001888.rn.1 RTA TC
          Rat Expression Set 230 1388155_at - -
          Rat Expression Set 230 1388155_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10745345 - -
          Affymetrix Rat Gene 1.0 ST Array 10940455 - -
          Affymetrix Rat Gene 1.0 ST Array 10813992 - -
          Rat Genome U34 Set X81448cds_at - -
          Rat Genome U34 Set rc_AI072634_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21434591 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline