Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01534598_m1
          See other KCTD9 GE Assays ›
          Gene Symbol:
          Kctd9
          Gene Name:
          potassium channel tetramerization domain containing 9
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 44411378 - 44439645 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01534598_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kctd9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108871.1 NP_001102341.1 11 - 12 1135 65
          XM_006252138.2 XP_006252200.1 10 - 11 971 65
          XM_006252139.2 XP_006252201.1 11 - 12 1368 65
          XM_006252141.2 XP_006252203.1 11 - 12 1438 65
          XM_006252142.2 XP_006252204.1 11 - 12 1052 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500000665.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s)

          protein homooligomerization

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P498522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443730 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13320 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106002 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78811 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122533 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P498522 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P498522 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P498522 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P498522 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P10399 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7604 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46986 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10781257 - -
          Affymetrix Rat Gene 1.0 ST Array 10781252 - -
          Affymetrix Rat Gene 1.0 ST Array 10781250 - -
          Affymetrix Rat Gene 1.0 ST Array 10781245 - -
          Affymetrix Rat Gene 1.0 ST Array 10781254 - -
          Affymetrix Rat Gene 1.0 ST Array 10781244 - -
          Affymetrix Rat Gene 1.0 ST Array 10781246 - -
          Affymetrix Rat Gene 1.0 ST Array 10781247 - -
          Affymetrix Rat Gene 1.0 ST Array 10781249 - -
          Affymetrix Rat Gene 1.0 ST Array 10781255 - -
          Affymetrix Rat Gene 1.0 ST Array 10781256 - -
          Affymetrix Rat Gene 1.0 ST Array 10781253 - -
          Affymetrix Rat Gene 1.0 ST Array 10781248 - -
          Affymetrix Rat Gene 1.0 ST Array 10781251 - -
          Affymetrix Rat Gene 1.1 ST Array 10781243 - -
          Rat Genome 230 2.0 Array 1374582_at - -
          Rat Genome 230 2.0 Array 1374582_PM_at - -
          Affymetrix HT Rat Focus Array 1374582_at - -
          Affymetrix HT Rat Focus Array 1374582_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1500004681.rn.1 TC1500000665.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500004680.rn.1 TC1500000665.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500004683.rn.1 TC1500000665.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500004682.rn.1 TC1500000665.rn.1 RTA TC
          Rat Expression Set 230 1374582_at - -
          Rat Expression Set 230 1374582_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10781243 - -
          Rat Genome U34 Set rc_AI175423_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21420340 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375166 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0