Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01534900_m1
          See other PINX1 GE Assays ›
          Gene Symbol:
          Pinx1
          Gene Name:
          PIN2/TERF1 interacting, telomerase inhibitor 1
          Gene Aliases:
          RGD1566025
          Chromosome Location:
          Chr.15: 47211682 - 47270343 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01534900_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pinx1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001083337.1 NP_001076806.1 6 - 7 473 86
          XM_006252181.2 XP_006252243.1 6 - 7 1376 86
          GenBank mRNA BC158553.1 - 6 - 7 572 86
          EF446165.1 - 6 - 7 473 86
          FQ232228.1 - 5 - 6 361 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500000708.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          telomere maintenance via telomerase
          mitotic metaphase plate congression
          telomerase RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P063818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814313 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70308 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137088 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72396 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65090 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40312 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18279 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55858 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164112 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P063818 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P063818 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P063818 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P563648 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 541 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56847 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10781017 - -
          Affymetrix Rat Gene 1.0 ST Array 10781015 - -
          Affymetrix Rat Gene 1.0 ST Array 10781018 - -
          Affymetrix Rat Gene 1.0 ST Array 10781021 - -
          Affymetrix Rat Gene 1.0 ST Array 10781019 - -
          Affymetrix Rat Gene 1.0 ST Array 10781016 - -
          Affymetrix Rat Gene 1.0 ST Array 10781020 - -
          Affymetrix Rat Gene 1.1 ST Array 10781014 - -
          Rat Genome 230 2.0 Array 1374854_PM_at - -
          Rat Genome 230 2.0 Array 1374854_at - -
          Affymetrix HT Rat Focus Array 1374854_PM_at - -
          Affymetrix HT Rat Focus Array 1374854_at - -
          Affymetrix Rat Clariom S Assay PSR1500004917.rn.1 TC1500000708.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500004918.rn.1 TC1500000708.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500004919.rn.1 TC1500000708.rn.1 RTA TC
          Rat Expression Set 230 1374854_PM_at - -
          Rat Expression Set 230 1374854_at - -
          Affymetrix Rat Gene 1.0 ST Array 10781014 - -
          Rat Genome U34 Set rc_AA924658_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20806213 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368485 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0