Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01536238_m1
          See other RBM12 GE Assays ›
          Gene Symbol:
          Rbm12 LOC100910990
          Gene Name:
          RNA binding motif protein 12
          copine-1-like
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.3: 151638167 - 151658445 on Build Rnor_6.0
          Chromosome Location:
          Chr.3: 152150143 - 152192954 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01536238_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rbm12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037657.1 NP_001032746.1 196 89
          XM_008762382.1 XP_008760604.1 2 - 3 256 89
          GenBank mRNA AY226099.1 - 196 89
          Gene Symbol: LOC100910990
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006235453.2 XP_006235515.1 2 - 3 224 89
          XM_008762412.1 XP_008760634.1 2 - 3 256 89
          XM_008762414.1 XP_008760636.1 1 - 2 2219 89

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AY226099.1, NM_001037657.1, XM_006235453.2, XM_008762382.1, XM_008762412.1, XM_008762414.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rbm12
          Click here to find siRNAs for Gene LOC100910990

          Gene Ontology Categories:

          Process(es)

          nucleotide binding
          nucleic acid binding
          RNA binding

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P250203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440624 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18387 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168304 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P250203 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P250203 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P250203 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P250203 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63096 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20584 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34477 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2358 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56099 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10851154 - -
          Affymetrix Rat Gene 1.0 ST Array 10851152 - -
          Affymetrix Rat Gene 1.0 ST Array 10851156 - -
          Affymetrix Rat Gene 1.0 ST Array 10851155 - -
          Affymetrix Rat Gene 1.0 ST Array 10851153 - -
          Affymetrix Rat Gene 1.0 ST Array 10851151 - -
          Affymetrix Rat Gene 1.1 ST Array 10851150 - -
          Rat Genome 230 2.0 Array 1377746_PM_at - -
          Rat Genome 230 2.0 Array 1391747_PM_at - -
          Rat Genome 230 2.0 Array 1377746_at - -
          Rat Genome 230 2.0 Array 1391747_at - -
          Affymetrix HT Rat Focus Array 1391747_PM_at - -
          Affymetrix HT Rat Focus Array 1377746_at - -
          Affymetrix HT Rat Focus Array 1391747_at - -
          Affymetrix HT Rat Focus Array 1377746_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300035101.rn.1 TC0300004566.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300035106.rn.1 TC0300004566.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300035103.rn.1 TC0300004566.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300035105.rn.1 TC0300004566.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300035104.rn.1 TC0300004566.rn.1 RTA TC
          Rat Expression Set 230 1391747_at - -
          Rat Expression Set 230 1377746_at - -
          Rat Expression Set 230 1377746_PM_at - -
          Rat Expression Set 230 1391747_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10851150 - -
          Rat Genome U34 Set rc_AI010418_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0