Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01537212_m1
          See other TBPL1 GE Assays ›
          Gene Symbol:
          Tbpl1
          Gene Name:
          TBP-like 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 23977072 - 24002512 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01537212_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tbpl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001127201.1 NP_001120673.1 4 - 5 533 77
          XM_006227721.2 XP_006227783.1 4 - 5 566 77
          XM_006227722.2 XP_006227784.1 4 - 5 490 77
          XM_008758627.1 XP_008756849.1 5 - 6 533 77
          GenBank mRNA CF109177.1 - 3 - 4 225 77
          FQ213058.1 - 3 - 4 185 77
          FQ220549.1 - 3 - 4 185 77
          FQ234587.1 - 4 - 5 551 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100000264.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          acrosome assembly
          dTTP biosynthetic process
          DNA-templated transcription, initiation
          spermatogenesis
          spermatid nucleus differentiation
          DNA binding

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P238642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262033 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3179 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58848 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100918 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P238642 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P238642 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P238642 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P238642 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10702316 - -
          Affymetrix Rat Gene 1.0 ST Array 10702312 - -
          Affymetrix Rat Gene 1.0 ST Array 10702313 - -
          Affymetrix Rat Gene 1.0 ST Array 10702314 - -
          Affymetrix Rat Gene 1.0 ST Array 10702310 - -
          Affymetrix Rat Gene 1.0 ST Array 10702311 - -
          Affymetrix Rat Gene 1.1 ST Array 10702309 - -
          Rat Genome 230 2.0 Array 1373242_PM_at - -
          Rat Genome 230 2.0 Array 1373242_at - -
          Affymetrix HT Rat Focus Array 1373242_PM_at - -
          Affymetrix HT Rat Focus Array 1373242_at - -
          Affymetrix Rat Clariom S Assay PSR0100001512.rn.1 TC0100000264.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100001513.rn.1 TC0100000264.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100001519.rn.1 TC0100000264.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100001520.rn.1 TC0100000264.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100001514.rn.1 TC0100000264.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100001523.rn.1 TC0100000264.rn.1 RTA TC
          Rat Expression Set 230 1373242_PM_at - -
          Rat Expression Set 230 1373242_at - -
          Affymetrix Rat Gene 1.0 ST Array 10702309 - -
          Rat Genome U34 Set rc_AI232431_at - -
          Rat Genome U34 Set rc_AI228170_at - -
          Rat Genome U34 Set rc_AI102800_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-ztd5s:80/100.66.134.6:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0