Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01537279_g1
          See other CTGF GE Assays ›
          Gene Symbol:
          Ctgf
          Gene Name:
          connective tissue growth factor
          Gene Aliases:
          CTGRP
          Chromosome Location:
          Chr.1: 21851657 - 21854773 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01537279_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ctgf
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022266.2 NP_071602.1 4 - 5 970 59
          GenBank mRNA AB023068.1 - 4 - 5 958 59
          AF079531.1 - 1 - 2 104 59
          AF120275.1 - 4 - 5 970 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100004176.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ctgf

          Gene Ontology Categories:

          Function(s) Process(es)

          cartilage condensation
          ossification
          angiogenesis
          regulation of cell growth
          tissue homeostasis
          positive regulation of protein phosphorylation
          cell adhesion
          cell-matrix adhesion
          integrin-mediated signaling pathway
          cell-cell signaling
          positive regulation of cell population proliferation
          fibroblast growth factor receptor signaling pathway
          response to glucose
          animal organ senescence
          positive regulation of gene expression
          negative regulation of gene expression
          positive regulation of cell death
          response to organic cyclic compound
          cell migration
          cell differentiation
          lung development
          regulation of chondrocyte differentiation
          response to estradiol
          positive regulation of collagen biosynthetic process
          response to anoxia
          intracellular signal transduction
          chondrocyte proliferation
          response to amino acid
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          response to peptide hormone
          positive regulation of cell differentiation
          positive regulation of JNK cascade
          positive regulation of cell activation
          response to mineralocorticoid
          positive regulation of stress fiber assembly
          cytosolic calcium ion transport
          positive regulation of cardiac muscle contraction
          negative regulation of cell death
          connective tissue development
          extracellular matrix constituent secretion
          positive regulation of G0 to G1 transition
          positive regulation of ERK1 and ERK2 cascade
          response to fatty acid
          DNA biosynthetic process
          reactive oxygen species metabolic process
          fibronectin binding
          integrin binding
          insulin-like growth factor binding
          growth factor activity
          heparin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P484738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763807 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P484738 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P484738 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P484738 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P484738 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82673 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10717236 - -
          Affymetrix Rat Gene 1.0 ST Array 10717238 - -
          Affymetrix Rat Gene 1.0 ST Array 10717239 - -
          Affymetrix Rat Gene 1.0 ST Array 10717237 - -
          Affymetrix Rat Gene 1.0 ST Array 10717235 - -
          Affymetrix Rat Gene 1.0 ST Array 10717234 - -
          Affymetrix Rat Gene 1.1 ST Array 10717233 - -
          Rat Genome 230 2.0 Array 1367631_at - -
          Rat Genome 230 2.0 Array 1367631_PM_at - -
          Affymetrix HT Rat Focus Array 1367631_at - -
          Affymetrix HT Rat Focus Array 1367631_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100033546.rn.1 TC0100004176.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100033545.rn.1 TC0100004176.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100033544.rn.1 TC0100004176.rn.1 RTA TC
          Rat Expression Set 230 1367631_at - -
          Rat Expression Set 230 1367631_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10717233 - -
          Rat Genome U34 Set rc_AA998395_f_at - -
          Rat Genome U34 Set rc_AI070318_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21024229 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364113 - -
          RatRef-12 Expression BeadChip ILMN_48683 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0