Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01537654_m1
          See other RNF217 GE Assays ›
          Gene Symbol:
          Rnf217
          Gene Name:
          ring finger protein 217
          Gene Aliases:
          Ibrdc1
          Chromosome Location:
          Chr.1: 28301960 - 28391582 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01537654_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnf217
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106204.1 NP_001099674.1 4 - 5 1030 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100000294.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Process(es)

          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P733175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78376 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53071 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32068 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23394 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131712 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125979 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56990 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105170 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136472 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18095 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167010 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44182 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P733175 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P733175 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P733175 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P733175 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71326 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89360 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44133 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10702350 - -
          Affymetrix Rat Gene 1.0 ST Array 10702351 - -
          Affymetrix Rat Gene 1.0 ST Array 10702343 - -
          Affymetrix Rat Gene 1.0 ST Array 10702346 - -
          Affymetrix Rat Gene 1.0 ST Array 10702345 - -
          Affymetrix Rat Gene 1.0 ST Array 10702344 - -
          Affymetrix Rat Gene 1.1 ST Array 10702342 - -
          Rat Genome 230 2.0 Array 1393047_PM_at - -
          Rat Genome 230 2.0 Array 1393047_at - -
          Affymetrix HT Rat Focus Array 1393047_PM_at - -
          Affymetrix HT Rat Focus Array 1393047_at - -
          Affymetrix Rat Clariom S Assay PSR0100001625.rn.1 TC0100000294.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100001624.rn.1 TC0100000294.rn.1 RTA TC
          Rat Expression Set 230 1393047_PM_at - -
          Rat Expression Set 230 1393047_at - -
          Affymetrix Rat Gene 1.0 ST Array 10702342 - -
          Rat Genome U34 Set rc_AI236773_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21192964 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365883 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline