Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01537740_m1
          See other OXSM GE Assays ›
          Gene Symbol:
          Oxsm
          Gene Name:
          3-oxoacyl-ACP synthase, mitochondrial
          Gene Aliases:
          RGD1311092
          Chromosome Location:
          Chr.15: 10462459 - 10466496 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01537740_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Oxsm
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001100508.1 NP_001093978.1 1 - 2 1226 67
          XM_006251711.1 XP_006251773.1 1 - 2 1009 67
          GenBank mRNA BC090025.1 - 1 - 2 162 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500000105.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          acyl-CoA metabolic process
          short-chain fatty acid biosynthetic process
          medium-chain fatty acid biosynthetic process
          3-oxoacyl-[acyl-carrier-protein] synthase activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P029461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786703 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P029461 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P029461 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P029461 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10779343 - -
          Affymetrix Rat Gene 1.0 ST Array 10779344 - -
          Affymetrix Rat Gene 1.1 ST Array 10779342 - -
          Rat Genome 230 2.0 Array 1391530_PM_a_at - -
          Rat Genome 230 2.0 Array 1391530_a_at - -
          Affymetrix HT Rat Focus Array 1391530_PM_a_at - -
          Affymetrix HT Rat Focus Array 1391530_a_at - -
          Affymetrix Rat Clariom S Assay PSR1500000932.rn.1 TC1500000105.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500000929.rn.1 TC1500000105.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500000926.rn.1 TC1500000105.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500000928.rn.1 TC1500000105.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500000933.rn.1 TC1500000105.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500000934.rn.1 TC1500000105.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500000924.rn.1 TC1500000105.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500000927.rn.1 TC1500000105.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500000925.rn.1 TC1500000105.rn.1 RTA TC
          Rat Expression Set 230 1391530_a_at - -
          Rat Expression Set 230 1391530_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10779342 - -
          Rat Genome U34 Set rc_AI137446_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21741475 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6f7b98477d-hxtzg:80/100.66.130.62:80.
          git-commit: 5163bf559e7ad0ebe9326a88094353fc6888890d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline