Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01538099_m1
          See other MYLK3 GE Assays ›
          Gene Symbol:
          Mylk3
          Gene Name:
          myosin light chain kinase 3
          Gene Aliases:
          MLCK, RGD1305801
          Chromosome Location:
          Chr.19: 27388163 - 27442451 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01538099_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mylk3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001110810.1 NP_001104280.1 8 - 9 2053 67
          XM_006255355.2 XP_006255417.1 8 - 9 1527 67
          GenBank mRNA EU236721.1 - 8 - 9 2053 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000372.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of vascular permeability involved in acute inflammatory response
          protein phosphorylation
          sarcomere organization
          sarcomerogenesis
          cardiac myofibril assembly
          positive regulation of sarcomere organization
          cellular response to interleukin-1
          calmodulin-dependent protein kinase activity
          myosin light chain kinase activity
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P100716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767952 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114463 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179726 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134050 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14078 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P100716 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P100716 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P100716 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P100716 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86630 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10806466 - -
          Affymetrix Rat Gene 1.0 ST Array 10806474 - -
          Affymetrix Rat Gene 1.0 ST Array 10806471 - -
          Affymetrix Rat Gene 1.0 ST Array 10806470 - -
          Affymetrix Rat Gene 1.0 ST Array 10806462 - -
          Affymetrix Rat Gene 1.0 ST Array 10806464 - -
          Affymetrix Rat Gene 1.0 ST Array 10806472 - -
          Affymetrix Rat Gene 1.0 ST Array 10806473 - -
          Affymetrix Rat Gene 1.0 ST Array 10806469 - -
          Affymetrix Rat Gene 1.0 ST Array 10806465 - -
          Affymetrix Rat Gene 1.0 ST Array 10806463 - -
          Affymetrix Rat Gene 1.0 ST Array 10806468 - -
          Affymetrix Rat Gene 1.0 ST Array 10806467 - -
          Affymetrix Rat Gene 1.1 ST Array 10806461 - -
          Rat Genome 230 2.0 Array 1382239_PM_at - -
          Rat Genome 230 2.0 Array 1382239_at - -
          Rat Genome 230 2.0 Array 1376789_PM_at - -
          Rat Genome 230 2.0 Array 1376789_at - -
          Affymetrix HT Rat Focus Array 1382239_at - -
          Affymetrix HT Rat Focus Array 1382239_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900003030.rn.1 TC1900000372.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900003032.rn.1 TC1900000372.rn.1 RTA TC
          Rat Expression Set 230 1382239_PM_at - -
          Rat Expression Set 230 1382239_at - -
          Affymetrix Rat Gene 1.0 ST Array 10806461 - -
          Rat Genome U34 Set rc_AI072534_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21005915 - -
          ABI Rat Genome Survey Microarray 21285898 - -
          ABI Rat Genome Survey Microarray 21333321 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355950 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0