Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01538252_m1
          See other SALL1 GE Assays ›
          Gene Symbol:
          Sall1
          Gene Name:
          spalt-like transcription factor 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 23387741 - 23405026 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01538252_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sall1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107415.2 NP_001100885.2 2 - 3 3533 148
          XM_006255259.2 XP_006255321.1 3 - 4 4067 148
          XM_006255260.2 XP_006255322.1 3 - 4 3658 148
          XM_008772401.1 XP_008770623.1 2 - 3 6539 148

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          ureteric bud development
          branching involved in ureteric bud morphogenesis
          kidney development
          neural tube closure
          ventricular septum development
          mesenchymal to epithelial transition involved in metanephros morphogenesis
          negative regulation of mesenchymal to epithelial transition involved in metanephros morphogenesis
          transcription by RNA polymerase II
          heart development
          biological_process
          gonad development
          olfactory nerve development
          olfactory bulb development
          olfactory bulb interneuron differentiation
          neural tube development
          pituitary gland development
          positive regulation of Wnt signaling pathway
          adrenal gland development
          inductive cell-cell signaling
          forelimb morphogenesis
          hindlimb morphogenesis
          outer ear morphogenesis
          embryonic digit morphogenesis
          positive regulation of neuron differentiation
          negative regulation of smoothened signaling pathway
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          embryonic digestive tract development
          limb development
          olfactory bulb mitral cell layer development
          kidney epithelium development
          ureteric bud invasion
          mesenchymal stem cell maintenance involved in metanephric nephron morphogenesis
          positive regulation of branching involved in ureteric bud morphogenesis
          regulation of neural precursor cell proliferation
          negative regulation of mesoderm development
          negative regulation of ectoderm development
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          molecular_function
          DNA binding
          chromatin binding
          transcription coactivator activity
          histone deacetylase activity
          beta-catenin binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P494591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930444 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120735 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174466 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P494591 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P494591 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P494591 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P494591 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10806316 - -
          Affymetrix Rat Gene 1.0 ST Array 10806317 - -
          Affymetrix Rat Gene 1.1 ST Array 10806314 - -
          Rat Genome 230 2.0 Array 1391194_at - -
          Rat Genome 230 2.0 Array 1391194_PM_at - -
          Affymetrix HT Rat Focus Array 1391194_at - -
          Affymetrix HT Rat Focus Array 1391194_PM_at - -
          Rat Expression Set 230 1391194_at - -
          Rat Expression Set 230 1391194_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10806314 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356064 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline