Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01538656_m1
          See other LOC100912163 GE Assays ›
          Gene Symbol:
          LOC100912163 Arid4b
          Gene Name:
          AT-rich interactive domain-containing protein 4B-like
          AT rich interactive domain 4B (Rbp1 like)
          Gene Aliases:
          -
          Bcaa
          Chromosome Location:
          Chr.17: 53629443 - 53741761 on Build Rnor_6.0
          Chromosome Location:
          Chr.17: 53831353 - 53955920 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01538656_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100912163
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008771836.1 XP_008770058.1 22 - 23 3943 90
          Gene Symbol: Arid4b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053421.1 NP_445873.1 21 - 22 3600 90
          XM_006254016.2 XP_006254078.1 23 - 24 4183 90
          XM_006254019.2 XP_006254081.1 22 - 23 3922 90
          XM_008771764.1 XP_008769986.1 23 - 24 4198 90
          XM_008771765.1 XP_008769987.1 23 - 24 4090 90
          XM_008771766.1 XP_008769988.1 22 - 23 3937 90
          GenBank mRNA AF245512.1 - 21 - 22 3600 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700001922.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC100912163
          Click here to find siRNAs for Gene Arid4b

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of gene expression by genetic imprinting
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription by RNA polymerase II
          spermatogenesis
          histone H4-K20 trimethylation
          histone H3-K9 trimethylation
          positive regulation of transcription by RNA polymerase II
          establishment of Sertoli cell barrier
          transcription cis-regulatory region binding
          DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12803 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P247966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40907 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136273 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28660 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62280 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103318 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47799 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8048 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100045 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75852 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163206 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12323 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35458 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155059 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167326 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P247966 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12803 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12803 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P247966 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P247966 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12803 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12803 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700695689 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700244762 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55121 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62820 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80127 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59214 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76870 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62285 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45108 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86493 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42742 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66271 - -
          Agilent Rat cDNA Microarray (G4105A) 700695689 - -
          Agilent Rat cDNA Microarray (G4105A) 700244762 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10798757 - -
          Affymetrix Rat Gene 1.0 ST Array 10798746 - -
          Affymetrix Rat Gene 1.0 ST Array 10798761 - -
          Affymetrix Rat Gene 1.0 ST Array 10798740 - -
          Affymetrix Rat Gene 1.0 ST Array 10798739 - -
          Affymetrix Rat Gene 1.0 ST Array 10798742 - -
          Affymetrix Rat Gene 1.0 ST Array 10798738 - -
          Affymetrix Rat Gene 1.0 ST Array 10798743 - -
          Affymetrix Rat Gene 1.0 ST Array 10798760 - -
          Affymetrix Rat Gene 1.0 ST Array 10798745 - -
          Affymetrix Rat Gene 1.0 ST Array 10798755 - -
          Affymetrix Rat Gene 1.0 ST Array 10798751 - -
          Affymetrix Rat Gene 1.0 ST Array 10798754 - -
          Affymetrix Rat Gene 1.0 ST Array 10798763 - -
          Affymetrix Rat Gene 1.0 ST Array 10798758 - -
          Affymetrix Rat Gene 1.0 ST Array 10798759 - -
          Affymetrix Rat Gene 1.0 ST Array 10798762 - -
          Affymetrix Rat Gene 1.0 ST Array 10798753 - -
          Affymetrix Rat Gene 1.0 ST Array 10798750 - -
          Affymetrix Rat Gene 1.0 ST Array 10798749 - -
          Affymetrix Rat Gene 1.0 ST Array 10798741 - -
          Affymetrix Rat Gene 1.0 ST Array 10798756 - -
          Affymetrix Rat Gene 1.0 ST Array 10798752 - -
          Affymetrix Rat Gene 1.1 ST Array 10798737 - -
          Rat Genome 230 2.0 Array 1368837_PM_at - -
          Rat Genome 230 2.0 Array 1368837_at - -
          Affymetrix HT Rat Focus Array 1368837_at - -
          Affymetrix HT Rat Focus Array 1368837_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1700012957.rn.1 TC1700001922.rn.1 RTA TC
          Rat Expression Set 230 1368837_at - -
          Rat Expression Set 230 1368837_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10798737 - -
          Rat Genome U34 Set rc_AA955478_at - -
          Rat Genome U34 Set rc_AI102488_at - -
          Rat Genome U34 Set rc_AI136605_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22346107 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354974 - -
          RatRef-12 Expression BeadChip ILMN_57097 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline