Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01636983_m1
          See other LOC103690035 GE Assays ›
          Gene Symbol:
          LOC103690035 Tgfbr1
          Gene Name:
          TGF-beta receptor type-1-like
          transforming growth factor, beta receptor 1
          Gene Aliases:
          -
          Alk5, Skr4, Tgfr-1, tbetaR-I
          Chromosome Location:
          Chr.5: 64565362 - 64585219 on Build Rnor_6.0
          Chromosome Location:
          Chr.5: 63056079 - 63124107 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01636983_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC103690035
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008763706.1 XP_008761928.1 1 - 2 1691 120
          Gene Symbol: Tgfbr1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006238030.2 XP_006238092.2 408 120
          GenBank mRNA L26110.1 - 363 120

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          angiogenesis
          response to hypoxia
          in utero embryonic development
          kidney development
          blastocyst development
          epithelial to mesenchymal transition
          negative regulation of endothelial cell proliferation
          positive regulation of endothelial cell proliferation
          lens development in camera-type eye
          regulation of transcription, DNA-templated
          protein phosphorylation
          apoptotic process
          signal transduction
          transmembrane receptor protein serine/threonine kinase signaling pathway
          transforming growth factor beta receptor signaling pathway
          heart development
          embryo implantation
          aging
          positive regulation of cell population proliferation
          germ cell migration
          male gonad development
          response to toxic substance
          post-embryonic development
          anterior/posterior pattern specification
          regulation of gene expression
          positive regulation of gene expression
          regulation of epithelial to mesenchymal transition
          positive regulation of pathway-restricted SMAD protein phosphorylation
          response to organic cyclic compound
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          collagen fibril organization
          positive regulation of cell growth
          lung development
          animal organ regeneration
          regulation of protein ubiquitination
          negative regulation of chondrocyte differentiation
          activin receptor signaling pathway
          response to prostaglandin E
          intracellular signal transduction
          regulation of growth
          endothelial cell activation
          negative regulation of apoptotic process
          regulation of protein binding
          endothelial cell migration
          response to estrogen
          negative regulation of endothelial cell differentiation
          positive regulation of transcription, DNA-templated
          protein autophosphorylation
          thymus development
          digestive tract development
          neuron fate commitment
          embryonic cranial skeleton morphogenesis
          skeletal system morphogenesis
          mesenchymal cell differentiation
          artery morphogenesis
          cell motility
          positive regulation of cellular component movement
          positive regulation of filopodium assembly
          response to electrical stimulus
          positive regulation of protein kinase B signaling
          parathyroid gland development
          roof of mouth development
          pharyngeal system development
          cardiac epithelial to mesenchymal transition
          pathway-restricted SMAD protein phosphorylation
          positive regulation of SMAD protein signal transduction
          response to cholesterol
          cellular response to transforming growth factor beta stimulus
          positive regulation of apoptotic signaling pathway
          negative regulation of extrinsic apoptotic signaling pathway
          protein kinase activity
          protein serine/threonine kinase activity
          transforming growth factor beta-activated receptor activity
          transforming growth factor beta receptor activity, type I
          type II transforming growth factor beta receptor binding
          ATP binding
          growth factor binding
          ubiquitin protein ligase binding
          protein-containing complex binding
          SMAD binding
          metal ion binding
          protein heterodimerization activity
          transforming growth factor beta binding
          I-SMAD binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline