Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01637803_m1
          See other SERPINA1 GE Assays ›
          Gene Symbol:
          Serpina1
          Gene Name:
          serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1
          Gene Aliases:
          AAT, Pi, Spi1
          Chromosome Location:
          Chr.6: 127610241 - 127632265 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01637803_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serpina1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022519.2 NP_071964.2 4 - 5 1150 114
          XM_006240456.1 XP_006240518.1 3 - 4 1007 114
          GenBank mRNA BC078824.1 - 4 - 5 1150 114
          D00675.1 - 3 - 4 953 114
          FQ209479.1 - 2 - 3 464 114
          FQ209494.1 - 2 - 3 467 114
          FQ209517.1 - 2 - 3 464 114
          FQ209677.1 - 2 - 3 464 114
          FQ209686.1 - 2 - 3 464 114
          FQ209716.1 - 2 - 3 465 114
          FQ209779.1 - 2 - 3 465 114
          FQ209814.1 - 2 - 3 465 114
          FQ209897.1 - 2 - 3 464 114
          FQ209914.1 - 2 - 3 464 114
          FQ210068.1 - 2 - 3 464 114
          FQ210655.1 - 3 - 4 943 114
          FQ210685.1 - 2 - 3 464 114
          FQ210775.1 - 2 - 3 464 114
          FQ210989.1 - 2 - 3 465 114
          FQ210996.1 - 2 - 3 465 114
          FQ211012.1 - 2 - 3 464 114
          FQ211042.1 - 2 - 3 464 114
          FQ211203.1 - 2 - 3 464 114
          FQ211353.1 - 2 - 3 464 114
          FQ211455.1 - 2 - 3 330 114
          FQ218226.1 - 2 - 3 464 114
          FQ218241.1 - 2 - 3 464 114
          FQ218261.1 - 2 - 3 464 114
          FQ218411.1 - 3 - 4 1083 114
          FQ218460.1 - 2 - 3 464 114
          FQ218555.1 - 2 - 3 464 114
          FQ218567.1 - 2 - 3 464 114
          FQ218654.1 - 2 - 3 464 114
          FQ218710.1 - 2 - 3 463 114
          FQ218855.1 - 2 - 3 464 114
          FQ218947.1 - 2 - 3 465 114
          FQ218955.1 - 2 - 3 465 114
          FQ218993.1 - 2 - 3 464 114
          FQ219224.1 - 2 - 3 464 114
          FQ219252.1 - 2 - 3 465 114
          FQ219268.1 - 2 - 3 464 114
          FQ219276.1 - 2 - 3 464 114
          FQ219383.1 - 2 - 3 464 114
          FQ219441.1 - 2 - 3 464 114
          FQ219621.1 - 2 - 3 464 114
          FQ219652.1 - 2 - 3 464 114
          FQ219732.1 - 2 - 3 464 114
          FQ219741.1 - 3 - 4 944 114
          FQ219765.1 - 2 - 3 464 114
          M32247.1 - 2 - 3 890 114
          X16273.1 - 2 - 3 336 114

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Serpina1

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          acute-phase response
          inflammatory response
          response to inorganic substance
          response to lead ion
          response to organic cyclic compound
          response to estradiol
          response to lipopolysaccharide
          response to methanol
          response to triglyceride
          response to cytokine
          response to chromate
          negative regulation of serine-type endopeptidase activity
          protease binding
          endopeptidase inhibitor activity
          serine-type endopeptidase inhibitor activity
          protein binding
          identical protein binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0