Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01642711_g1
          See other DDX39A GE Assays ›
          Gene Symbol:
          Ddx39a
          Gene Name:
          DEAD (Asp-Glu-Ala-Asp) box polypeptide 39A
          Gene Aliases:
          Ddx39, Ddxl
          Chromosome Location:
          Chr.19: 24846535 - 24854886 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01642711_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ddx39a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053563.2 NP_446015.2 9 - 10 1420 133
          XM_006255314.2 XP_006255376.1 9 - 10 1320 133
          GenBank mRNA AF063447.1 - 9 - 10 1216 133
          BC086328.1 - 9 - 10 1420 133
          FQ231639.1 - 9 - 10 1326 133

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000305.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ddx39a

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splicing, via spliceosome
          mRNA export from nucleus
          RNA binding
          helicase activity
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P149412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27183 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134224 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P149412 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P149412 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P149412 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P149412 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700176125 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75295 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24155 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64920 - -
          Agilent Rat cDNA Microarray (G4105A) 700176125 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10810272 - -
          Affymetrix Rat Gene 1.0 ST Array 10810268 - -
          Affymetrix Rat Gene 1.0 ST Array 10810275 - -
          Affymetrix Rat Gene 1.0 ST Array 10810266 - -
          Affymetrix Rat Gene 1.0 ST Array 10810269 - -
          Affymetrix Rat Gene 1.0 ST Array 10810276 - -
          Affymetrix Rat Gene 1.0 ST Array 10810273 - -
          Affymetrix Rat Gene 1.0 ST Array 10810267 - -
          Affymetrix Rat Gene 1.0 ST Array 10810270 - -
          Affymetrix Rat Gene 1.0 ST Array 10810265 - -
          Affymetrix Rat Gene 1.0 ST Array 10810271 - -
          Affymetrix Rat Gene 1.0 ST Array 10810274 - -
          Affymetrix Rat Gene 1.1 ST Array 10810264 - -
          Rat Genome 230 2.0 Array 1387048_at - -
          Rat Genome 230 2.0 Array 1387048_PM_at - -
          Affymetrix HT Rat Focus Array 1387048_PM_at - -
          Affymetrix HT Rat Focus Array 1387048_at - -
          Affymetrix Rat Clariom S Assay PSR1900002298.rn.1 TC1900000305.rn.1 RTA TC
          Rat Expression Set 230 1387048_PM_at - -
          Rat Expression Set 230 1387048_at - -
          Affymetrix Rat Gene 1.0 ST Array 10810264 - -
          Rat Genome U34 Set AF063447_at - -
          Rat Genome U34 Set rc_AI233796_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21614895 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367954 - -
          RatRef-12 Expression BeadChip ILMN_52155 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0