Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01746979_m1
          See other FRZB GE Assays ›
          Gene Symbol:
          Frzb
          Gene Name:
          frizzled-related protein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 67635604 - 67668772 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01746979_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Frzb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001100527.1 NP_001093997.1 6 - 7 1243 63
          XM_006234429.2 XP_006234491.1 5 - 6 1667 63
          GenBank mRNA BC098777.1 - 3 - 4 341 63
          BC166428.1 - 6 - 7 1243 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300003408.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          negative regulation of cell population proliferation
          negative regulation of cell development
          neural crest cell differentiation
          negative regulation of Wnt signaling pathway
          negative regulation of cell growth
          non-canonical Wnt signaling pathway
          inner ear morphogenesis
          positive regulation of apoptotic process
          positive regulation of fat cell differentiation
          convergent extension involved in organogenesis
          negative regulation of cartilage development
          somite development
          negative regulation of hepatocyte differentiation
          negative regulation of canonical Wnt signaling pathway
          cochlea morphogenesis
          molecular_function
          G protein-coupled receptor activity
          Wnt-protein binding
          Wnt-activated receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P152790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102196 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50285 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161352 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P152790 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P152790 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P152790 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P443220 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92172 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10846746 - -
          Affymetrix Rat Gene 1.0 ST Array 10846742 - -
          Affymetrix Rat Gene 1.0 ST Array 10846745 - -
          Affymetrix Rat Gene 1.0 ST Array 10846741 - -
          Affymetrix Rat Gene 1.0 ST Array 10846743 - -
          Affymetrix Rat Gene 1.0 ST Array 10846744 - -
          Affymetrix Rat Gene 1.1 ST Array 10846740 - -
          Rat Genome 230 2.0 Array 1373615_PM_at - -
          Rat Genome 230 2.0 Array 1375961_at - -
          Rat Genome 230 2.0 Array 1375961_PM_at - -
          Rat Genome 230 2.0 Array 1373615_at - -
          Affymetrix HT Rat Focus Array 1375961_PM_at - -
          Affymetrix HT Rat Focus Array 1375961_at - -
          Affymetrix HT Rat Focus Array 1373615_at - -
          Affymetrix HT Rat Focus Array 1373615_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300026788.rn.1 TC0300003408.rn.1 RTA TC
          Rat Expression Set 230 1375961_at - -
          Rat Expression Set 230 1373615_at - -
          Rat Expression Set 230 1373615_PM_at - -
          Rat Expression Set 230 1375961_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10846740 - -
          Rat Genome U34 Set rc_AI227737_at - -
          Rat Genome U34 Set rc_AI072892_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21235166 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366268 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix